Skip to Content

38152

N-[Nα,Nα-Bis(carboxymethyl)-L-lysine]-12-mercaptododecanamide

≥90.0% (TLC)

Synonym(s):

N2,N2-Bis(carboxymethyl)-N6-(12-mercapto-1-oxododecyl)-L-lysine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H40N2O7S
CAS Number:
Molecular Weight:
476.63
UNSPSC Code:
12352209
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
9676381
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90.0% (TLC)

mp

128-134 °C

storage temp.

2-8°C

SMILES string

OC(=O)CN(CC(O)=O)[C@@H](CCCCNC(=O)CCCCCCCCCCCS)C(O)=O

InChI

1S/C22H40N2O7S/c25-19(13-8-6-4-2-1-3-5-7-11-15-32)23-14-10-9-12-18(22(30)31)24(16-20(26)27)17-21(28)29/h18,32H,1-17H2,(H,23,25)(H,26,27)(H,28,29)(H,30,31)/t18-/m0/s1

InChI key

DFNBCRSGLOPDAH-SFHVURJKSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library