Skip to Content
Merck
All Photos(1)

Key Documents

347M-1

Sigma-Aldrich

Vimentin (V9) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

V9, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (347M-14)
vial of 0.5 mL concentrate (347M-15)
bottle of 1.0 mL predilute (347M-17)
vial of 1.0 mL concentrate (347M-16)
bottle of 7.0 mL predilute (347M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgGκ

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... VIM(7431)

General description

Anti-vimentin is of limited value as a diagnostic tool; however, when used in combination with other antibodies (in panels) it is useful for the subclassification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-vimentin. This antibody recognizes a 57 kD intermediate filament. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain vimentin, e.g. hepatoma and seminoma. Anti-vimentin is also useful as a tissue process control reagent.

Quality


IVD

IVD

IVD

RUO

Linkage

Vimentin Positive Control Slides, Product No. 347S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ben-Ze'ev
The Journal of cell biology, 99(4 Pt 1), 1424-1433 (1984-10-01)
The expression of cytokeratins and vimentin was investigated in Madin-Darby bovine epithelial cells (MDBK) in culture under conditions of varied cell spreading and cell-cell contact. When extensive cell-cell contact was achieved by seeding cells at high density in monolayer, or
E B Lane et al.
Nature, 303(5919), 701-704 (1983-06-23)
Of the five classes of intermediate filaments found in vertebrate tissues, the cytokeratins are considered unique to epithelial tissues, while vimentin is expressed by endothelial and mesenchymal cells. In neither case is the precise function of the filament system known.
M Leader et al.
Histopathology, 11(1), 63-72 (1987-01-01)
In this study we examined 198 sarcomas, 38 carcinomas, 13 'tumours with a spindle cell component' and 22 malignant melanomas with a commercial monoclonal vimentin antibody. All histopathological material was formalin fixed and paraffin embedded. The results show this antibody
I Takeyoshi et al.
Hepato-gastroenterology, 47(36), 1611-1614 (2001-01-10)
Carcinosarcomas are rare tumors, which are usually composed of carcinomatous areas close to or intermixed with sarcomatous components. Only 6 cases of carcinosarcoma of the colon have been reported in the English literature previously. We describe the 7th case, an
F R Davey et al.
The American journal of pathology, 129(1), 54-63 (1987-10-01)
The use of monoclonal and polyclonal antibodies for the immunophenotyping of non-Hodgkin's lymphomas in paraffin-embedded tissue has been limited by the fact that most antigens on lymphoid cells are denatured by histologic fixation, dehydration, and embedment. In this article the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service