Skip to Content
Merck
All Photos(1)

Key Documents

31395

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

Mr ~500,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

biological source

(Leuconostoc spp.)

form

powder

mol wt

Mr ~500,000

composition

Sulfur Content, 16.0-19.0% (dry basis)

manufacturer/tradename

EDQM

useful pH range

6.0-8.0 (10 g/L)

solubility

H2O: freely soluble 50 mg/mL, clear

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤1000 mg/kg
Mg: ≤100 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Dextran sulfate sodium salt from Leuconostoc sp. was used as a test compound for studying the heparin and dextran sulphate interaction with alkali ions. It was used for releasing DNA from DNA-histone complexes.

Biochem/physiol Actions

Dextran is a mixture of glucose polymers of varying size and geometry. Dextran in the form of dextran sulfate acts as an anticoagulant. Dextran interferes with blood clot formation by acting on blood coagulation factors and the function of platelets. It prolongs the time required for formation of the clot and decreases the maximum clot strength.The disturbance of homeostasis by dextran may cause increased bleeding, colitis and kidney failure.

Preparation Note

prepared from dextran with Mr ~500,000

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heparin and dextran sulfate antagonize PGI2 inhibition of platelet aggregation.
A Eldor et al.
Thrombosis research, 16(5-6), 617-628 (1979-01-01)
Prevention of venous thromboembolism.
G P Clagett et al.
Chest, 108(4 Suppl), 312S-334S (1995-10-01)
Lisa Nebelin Hvidt et al.
Danish medical journal, 59(11), A4531-A4531 (2012-11-23)
Synthetic colloids are frequently used in fluid resuscitation of septic patients. Despite this, little is known about the potential side effects including the risk of renal failure and bleeding. As practice has changed, we performed a before-and-after study of fluid
Simon Finfer et al.
The New England journal of medicine, 350(22), 2247-2256 (2004-05-28)
It remains uncertain whether the choice of resuscitation fluid for patients in intensive care units (ICUs) affects survival. We conducted a multicenter, randomized, double-blind trial to compare the effect of fluid resuscitation with albumin or saline on mortality in a
Se Young Choi et al.
Journal of biomedicine & biotechnology, 2010, 943516-943516 (2010-03-20)
A total of 28 male BALB/c mice (average weight 20.7 +/- 1.6 g) were divided into 4 treatment groups and fed a commercial diet (A), a commercial diet + induced colitis by dextran sodium sulfate (DSS) (B), Inonotus obliquus (IO)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service