Skip to Content
Merck

Skip To

30633

Sodium 1-decanesulfonate

≥99.0% (T)

Synonym(s):

1-Decanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)8CH2SO3Na
CAS Number:
Molecular Weight:
244.33
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
236-525-9
Beilstein/REAXYS Number:
3918920
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Segment

assay

≥99.0% (T)

mol wt

244.33 g/mol

technique(s)

electrophoresis: suitable

mp

>300 °C (lit.)

solubility

H2O: 0.05 g/mL, clear

SMILES string

[Na+].CCCCCCCCCCS([O-])(=O)=O

InChI

1S/C10H22O3S.Na/c1-2-3-4-5-6-7-8-9-10-14(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1

InChI key

AIMUHNZKNFEZSN-UHFFFAOYSA-M

General description

Sodium 1-decanesulfonate is an ion-associating reagent.

Application

Ion-associating reagent for HPLC, including analyses of peptides and proteins.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
30631D3412106437
assay

≥99.0% (T)

assay

≥99.0% (T)

assay

~98%

assay

≥99%

technique(s)

electrophoresis: suitable

technique(s)

ion pair chromatography: suitable

technique(s)

LC/MS: suitable

technique(s)

-

mp

>300 °C (lit.)

mp

>300 °C (lit.)

mp

>300 °C (lit.)

mp

>300 °C (lit.)

description

anionic

description

anionic

description

anionic

description

-

mol wt

244.33 g/mol

mol wt

244.33 g/mol

mol wt

244.33 g/mol

mol wt

-

solubility

H2O: 0.05 g/mL, clear

solubility

-

solubility

-

solubility

H2O: 25 mg/mL


Still not finding the right product?

Explore all of our products under Sodium 1-decanesulfonate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters