Skip to Content
Merck

3010

Transcreener® ADP2 FP Assay

Synonym(s):

Transcreener assay

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41122100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

storage temp.

−20°C

General description

The Transcreener® HTS Assay platform overcomes the need for time-consuming, one-off assay development for individual members within a group transfer enzyme family by utilizing a single set of assay reagents that detect an invariant product. The generic nature of the Transcreener® HTS Assay platform eliminates delays involved in assay development for new HTS targets, and greatly simplifies compound and inhibitor profiling across multiple target families.

The Transcreener® ADP2 FP Assay is a far-red, competitive fluorescence polarization (FP) assay based on the detection of ADP and therefore is compatible with any enzyme class that produces ADP, including protein, lipid, and carbohydrate kinases, ATPases, DNA helicases, carboxylases and glutamine synthetase. The Transcreener® ADP2 Assay is a simple one step homogenous detection assay, and is extremely flexible with regard to ATP concentration (0.1 to 1,000 μM ATP). The assay provides excellent signal at low substrate conversion, with a Z′ = 0.7 and = 85 polarization shift (mP) at 10% ATP conversion using 1 μM ATP.

The Transcreener® ADP2 FP Assay was developed to follow the progress of any enzyme that produces ADP. The Transcreener® ADP Detection Mixture comprises an ADP Alexa633 Tracer bound to an ADP2 Antibody. The tracer is displaced by ADP, the invariant product generated during the enzyme reaction.The displaced tracer freely rotates leading to a decrease in fluorescence polarization. The assay uses a far red tracer to minimize interference from fluorescent compounds and light scattering.

View full Transcreener® product list

Physical form

Kit with buffered aqueous solutions

Preparation Note

3010-1K = 1,000 assay, 384-well
3010-10K = 10,000 assay, 384-well

Legal Information

Transcreener is a registered trademark of BellBrook Labs


signalword

Warning

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

target_organs

Respiratory Tract

pictograms

Health hazardExclamation mark

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT RE 2 Inhalation



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library