Skip to Content
Merck
All Photos(1)

Key Documents

28616

Sigma-Aldrich

Atto 425-Biotin

BioReagent, suitable for fluorescence, ≥95.0% (HPCE)

Synonym(s):

Biotin-Atto 425

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H53N5O7S
Molecular Weight:
711.91
MDL number:
UNSPSC Code:
12352108
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥95.0% (HPCE)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 437 nm; λem 483 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCOC(=O)C1=Cc2cc3C(C)CC(C)(C)N(CCCC(=O)NCCCCCNC(=O)CCCCC4SCC5NC(=O)NC45)c3cc2OC1=O

InChI

1S/C37H53N5O7S/c1-5-48-34(45)26-19-24-18-25-23(2)21-37(3,4)42(28(25)20-29(24)49-35(26)46)17-11-14-32(44)39-16-10-6-9-15-38-31(43)13-8-7-12-30-33-27(22-50-30)40-36(47)41-33/h18-20,23,27,30,33H,5-17,21-22H2,1-4H3,(H,38,43)(H,39,44)(H2,40,41,47)

InChI key

LPVYZZMUDCZDQO-UHFFFAOYSA-N

General description

Atto 425 is a new label with high quantum yield (0.90) as well as sufficient stoke′s shift (excitation maximum 436 nm, emission maximum 484 nm). It is optimized for excitation with argon laser, and is characterized by high photostability. It does nearly not show cis-trans-isomerization, which limits brightness and reproducibility for many other dyes.

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 425 biotin is suitable for fluorescence labeling of avidin/streptavidin conjugated molecules.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunohistochemical methods for the demonstration of tumor markers.
H Denk
Current topics in pathology. Ergebnisse der Pathologie, 77, 47-69 (1987-01-01)
J Oertel et al.
Journal of cancer research and clinical oncology, 126(8), 425-440 (2000-08-29)
Immunocytochemical investigations of fixed cells are used to enhance diagnostic accuracy in haematology and oncology. The alkaline-phosphatase/anti(alkaline phosphatase) technique, immunoperoxidase and the avidin-biotin technique are the most important methods in immunocytochemistry. Tyramide-enhanced immunostaining is a new powerful technique. Peripheral blood
Katrin G Heinze et al.
Biophysical journal, 86(1 Pt 1), 506-516 (2003-12-26)
Confocal fluorescence spectroscopy is a versatile method for studying dynamics and interactions of biomolecules in their native environment with minimal interference with the observed system. Analyzing coincident fluctuations induced by single molecule movement in spectrally distinct detection channels, dual-color fluorescence
Triple FRET: a tool for studying long-range molecular interactions.
Elke Haustein et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 4(7), 745-748 (2003-08-07)
Anna I Sulatskaya et al.
The journal of physical chemistry. B, 116(8), 2538-2544 (2012-01-25)
Benzothiazole dye thioflavin T (ThT) is a sensitive probe for amyloid fibril detection. The ThT probing is based on its unique ability to form highly fluorescent complexes with amyloid and amyloid-like fibrils. In this work we propose an approach of

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service