Skip to Content
Merck
All Photos(1)

Key Documents

27597

Sigma-Aldrich

Coenzyme Q9

≥96.0% (HPLC)

Synonym(s):

Q-9, Ubiquinone-45, Ubiquinone-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H82O4
CAS Number:
Molecular Weight:
795.23
Beilstein:
1900081
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

Assay

≥96.0% (HPLC)

form

powder

application(s)

cell analysis

storage temp.

−20°C

SMILES string

COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C)=C(C)C1=O

InChI

1S/C54H82O4/c1-40(2)22-14-23-41(3)24-15-25-42(4)26-16-27-43(5)28-17-29-44(6)30-18-31-45(7)32-19-33-46(8)34-20-35-47(9)36-21-37-48(10)38-39-50-49(11)51(55)53(57-12)54(58-13)52(50)56/h22,24,26,28,30,32,34,36,38H,14-21,23,25,27,29,31,33,35,37,39H2,1-13H3/b41-24+,42-26+,43-28+,44-30+,45-32+,46-34+,47-36+,48-38+

InChI key

UUGXJSBPSRROMU-WJNLUYJISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Coenzyme Q9 (Ubiquinone-9; Ubi-9) is a 9 isoprenyl group member of a family of ubiquinones that share a quinine chemical group but differ in the number of isoprenyl chemical subunits in their tail. The CoQ compounds are lipid soluble components of cell membranes.[1]

Application

Coenzyme Q9 (Ubiquinone-9; Ubi-9) may be used in the study of ubiquinones in vivo and in vitro. Coenzyme Q9 may be used as a reference compound in analytic procedures that monitor specific ubiquinone levels under various physiological and pathological conditions and in food-stocks.[2]
Coenzyme Q9 has been used to investigate its interference in the detection of coenzyme Q10 (CoQ10) by fluorescent spectrophotometer (FS-ECA).[3] It has also been used as an analyte in chromatographic analysis of different biological samples that correspond to different genetic conditions.[4]

Biochem/physiol Actions

The coenzyme Q (CoQ) can perform multiple functions such as electron and proton transport.[1] CoQ9 performs similar functions to CoQ10 as the native CoQ in Caenorhabditis elegans. CoQ9 as well as other CoQ molecules may have an impact on cell life-span and these molecules are seeing increase usage in such studies.[5]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

An efficient electron and proton translocator in liposome systems;[6] Prevents the appearance of adriamycin®-associated cardiotoxic phenomena in mice and rabbits.[7]

Legal Information

Adriamycin is a registered trademark of RDF Pharmacia & Upjohn S.P.A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

13 C-isotope-based protocol for prenyl lipid metabolic analysis in zebrafish embryos
Mugoni V, et al.
Nature Protocols, 8(12), 2337-2337 (2013)
Yuzuru Akamatsu
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 88(10), 536-553 (2012-12-12)
By using "our devised up-to-the-second technique" over 30 years ago, we succeeded in the first isolation in the world of the three different kinds of mammalian cell mutants defective in the biosynthesis on each of phosphatidylserine (PS), cardiolipin (CL) and
Carmen Venegas et al.
Journal of agricultural and food chemistry, 59(22), 12102-12108 (2011-10-20)
Virgin argan oil possesses high antioxidant capacity (AC), which may be partially explained by its high content in antioxidant molecules such as polyphenols and tocopherols. However, the content in other antioxidant molecules, for example, coenzyme Q10 (CoQ(10)), coenzyme Q9 (CoQ(9))
Adriamycin associated cardiotoxicity: research on prevention with coenzyme Q.
C Bertazzoli et al.
Pharmacological research communications, 9(3), 235-250 (1977-03-01)
G. Hauska
Dev. Bioenerg. Biomembr., 1, 177-177 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service