Skip to Content
Merck
All Photos(1)

Key Documents

21884

Sigma-Aldrich

5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate

BioReagent, suitable for fluorescence, ≥85% (HPCE)

Synonym(s):

5&6DH-DA, CDFDA, Carboxy-DCFDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H14Cl2O9
CAS Number:
Molecular Weight:
529.28
Beilstein:
7505106
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥85% (HPCE)

mp

210 °C (dec.) (lit.)

solubility

DMF: soluble
DMSO: soluble
methanol: soluble

fluorescence

λex 470 nm; λem 529 nm in 0.1 M Tris pH 8.0 (esterase)

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)Oc1cc2Oc3cc(OC(C)=O)c(Cl)cc3C4(OC(=O)c5cc(ccc45)C(O)=O)c2cc1Cl.CC(=O)Oc6cc7Oc8cc(OC(C)=O)c(Cl)cc8C9(OC(=O)c%10ccc(cc9%10)C(O)=O)c7cc6Cl

InChI

1S/2C25H14Cl2O9/c1-10(28)33-21-8-19-15(6-17(21)26)25(14-4-3-12(23(30)31)5-13(14)24(32)36-25)16-7-18(27)22(34-11(2)29)9-20(16)35-19;1-10(28)33-21-8-19-15(6-17(21)26)25(14-5-12(23(30)31)3-4-13(14)24(32)36-25)16-7-18(27)22(34-11(2)29)9-20(16)35-19/h2*3-9H,1-2H3,(H,30,31)

InChI key

LZMPJBOMBCYYMP-UHFFFAOYSA-N

General description

5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate or CDFDA is a fluorescent oxidant sensing probe. It is an intracellular probe added to the cells as a pro-substrate, which upon hydrolysis by the esterase enzyme produces the fluorescent product called CDF.

Application

5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate or Carboxy-DCFDA is used as a cell permeate probe to detect intracellular reactive oxygen species (ROS). The anionic fluorescent dye, 5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate is suitable for studying Multi-Drug Resistance (MDR) by target cells.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

Excmax: 504 nm (Lit.)

Other Notes

Intracellular esterase substrate with low sensitivity of fluorescence to intracellar pH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C K Raymond et al.
The Journal of cell biology, 111(3), 877-892 (1990-09-01)
vps3 mutants of the yeast Saccharomyces cerevisiae are impaired in the sorting of newly synthesized soluble vacuolar proteins and in the acidification of the vacuole (Rothman, J. H., and T. H. Stevens. Cell. 47:1041-1051; Rothman, J. H., C. T. Yamashiro
Tassarut Chaisit et al.
European journal of pharmacology, 795, 50-57 (2016-12-06)
Rhinacanthin-C is a major bioactive naphthoquinone ester found in Rhinacanthus nasutus Kurz (Acanthaceae). This compound has potential therapeutic value as an anticancer and antiviral agent. In this study, we investigated an enhancement effect of rhinacanthin-C on doxorubicin cytotoxicity in human
Guillermo Nicolás Tocchetti et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 122, 205-213 (2018-07-10)
Multidrug resistance-associated protein 2 (MRP2) plays a key role in hepatic and intestinal disposition of endo- and xenobiotics. Several therapeutic agents modulate MRP2 activity resulting in pharmacological interactions. Nomegestrol acetate (NMGA) is a progestogen increasingly used in contraceptive formulations. The
Surgical treatment of the symptomatic juvenile flexible flatfoot condition.
G L Dockery
Clinics in podiatric medicine and surgery, 4(1), 99-117 (1987-01-01)
Yu-Fei Yao et al.
Life sciences, 180, 151-159 (2016-12-14)
(-)-Epigallocatechin-3-gallate (EGCG), a major green tea polyphenol compound, plays an important role in the prevention of cardiovascular disease and cancer. The present study aimed to investigate the effects of EGCG on doxorubicin (DOX)-induced cardiotoxicity in Sarcoma 180 (S180) tumor-bearing mice.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service