Skip to Content
Merck
All Photos(1)

Documents

18842

Sigma-Aldrich

Octyl-β-D-glucopyranoside 100 mM solution

Synonym(s):

Octyl-β-D-glucopyranoside solution, Detergent Screening Solution 03/Kit-No 66317

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.32

description

non-ionic

Quality Level

form

liquid

technique(s)

cell culture | insect: suitable
flow cytometry: suitable

storage temp.

2-8°C

SMILES string

CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaoqun Yao et al.
Proteomics. Clinical applications, 4(1), 4-16 (2010-12-08)
About two million new cases of leishmaniasis with 50 000 associated deaths occur worldwide each year. Promastigotes of the causative Leishmania spp. develop from the procyclic stage to the highly virulent metacyclic stage within the sand fly vector. We hypothesized
Christine Ménager et al.
Langmuir : the ACS journal of surfaces and colloids, 26(19), 15453-15463 (2010-09-10)
The present study deals with the morphological modifications of giant dioleoyl phosphatidylcholine vesicles (DOPC GUVs) induced by the nonionic surfactant n-octyl β,D-glucopyranoside at sublytic levels, i.e., in the first steps of the vesicle-to-micelle transition process, when surfactant inserts into the
J D Doran et al.
Journal of pharmaceutical sciences, 101(1), 92-101 (2011-09-17)
Membrane proteins are attractive therapeutic targets, however the presence of detergents complicates biophysical binding measurements. Difficulties in determining quantitative dissociation constants for problematic membrane proteins were addressed by combining analytical ultracentrifugation and classical light scattering techniques. Validation of the algorithm
Zahrabatoul Mosapour Kotena et al.
Journal of molecular modeling, 19(2), 589-599 (2012-09-14)
Density functional theory calculations on two glycosides, namely, n-octyl-β-D-glucopyranoside (C(8)O-β-Glc) and n-octyl-β-D-galactopyranoside (C(8)O-β-Gal) were performed for geometry optimization at the B3LYP/6-31G level. Both molecules are stereoisomers (epimers) differing only in the orientation of the hydroxyl group at the C4 position.
Kerry K Karukstis et al.
The journal of physical chemistry. B, 116(12), 3816-3822 (2012-03-06)
When mixed with water, n-octyl β-D-glucoside forms self-assembled nanostructures, several of which are liquid crystalline and all of which depend on the water/glucoside ratio and temperature. For practical use of these phases, a detailed understanding of the conditions under which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service