Skip to Content
Merck
All Photos(1)

Key Documents

16609

Sigma-Aldrich

1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester

≥95% (HPLC)

Synonym(s):

5,5′-Dimethyl-BAPTA-AM, MAPTAM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H44N2O18
CAS Number:
Molecular Weight:
792.74
Beilstein:
8181451
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(C)cc1OCCOc2cc(C)ccc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O

InChI

1S/C36H44N2O18/c1-23-7-9-29(37(15-33(43)53-19-49-25(3)39)16-34(44)54-20-50-26(4)40)31(13-23)47-11-12-48-32-14-24(2)8-10-30(32)38(17-35(45)55-21-51-27(5)41)18-36(46)56-22-52-28(6)42/h7-10,13-14H,11-12,15-22H2,1-6H3

InChI key

HEOJVQZWOLQUDR-UHFFFAOYSA-N

Other Notes

Membrane-permeable improved derivative of BAPTA; chelators for intracellular calcium incorporated non-disruptively into cells

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian John et al.
Frontiers in molecular neuroscience, 10, 171-171 (2017-07-01)
Glioblastoma multiforme (GBM) is a highly aggressive type of brain tumor with an extremely poor prognosis. Recent evidences have shown that the "biomechanical imbalances" induced in GBM patient-derived glioblastoma cells (GC) and in vivo via the administration of synthetic small
T Tiffert et al.
The Journal of physiology, 505 ( Pt 2), 403-410 (1998-01-10)
1. A recently developed method of measuring cytoplasmic Ca2+ buffering in intact red cells was applied to re-evaluate the intracellular Ca2+ binding properties of the Ca2+ chelators benz2 and BAPTA. Incorporation of the free chelators was accomplished by incubating the
J M Dubinsky
Neuroscience letters, 150(2), 129-132 (1993-02-19)
In an attempt to probe the relationship between excitotoxicity and increases in intracellular calcium ([Ca2+]i), BAPTA-AM and its analogs were applied to cultured hippocampal neurons. Chelation of [Ca2+]i depressed and prolonged transient responses to glutamate and did not effect elevation
Nida Arif et al.
The EMBO journal, 40(9), e106113-e106113 (2021-02-20)
Leukocyte extravasation is an essential step during the immune response and requires the destabilization of endothelial junctions. We have shown previously that this process depends in vivo on the dephosphorylation of VE-cadherin-Y731. Here, we reveal the underlying mechanism. Leukocyte-induced stimulation
Shah Alam et al.
Cells, 9(10) (2020-10-02)
We have shown that sphingosine 1-phosphate (S1P) generated by sphingosine kinase 2 (SK2) is toxic in neurons lacking S1P-lyase (SGPL1), the enzyme that catalyzes its irreversible cleavage. Interestingly, patients harboring mutations in the gene encoding this enzyme (SGPL1) often present

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service