Skip to Content
Merck
All Photos(1)

Key Documents

14370

Sigma-Aldrich

Bilirubin

purum, ≥95.0% (UV)

Synonym(s):

Bile pigment, Hemetoidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H36N4O6
CAS Number:
Molecular Weight:
584.66
Beilstein:
74376
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25

grade

purum

Assay

≥95.0% (UV)

form

powder

ign. residue

≤0.5%

SMILES string

CC1=C(C=C)\C(NC1=O)=C\c2[nH]c(Cc3[nH]c(\C=C4/NC(=O)C(C=C)=C4C)c(C)c3CCC(O)=O)c(CCC(O)=O)c2C

InChI

1S/C33H36N4O6/c1-7-20-19(6)32(42)37-27(20)14-25-18(5)23(10-12-31(40)41)29(35-25)15-28-22(9-11-30(38)39)17(4)24(34-28)13-26-16(3)21(8-2)33(43)36-26/h7-8,13-14,34-35H,1-2,9-12,15H2,3-6H3,(H,36,43)(H,37,42)(H,38,39)(H,40,41)/b26-13-,27-14-

InChI key

BPYKTIZUTYGOLE-IFADSCNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bilirubin, a heme catabolism end-product is produced by the reduction of its metabolic precursor biliverdin by the action of enzyme biliverdin reductase.

Application

Bilirubin has been used as an interfering substance for in vitro interference and stability tests for myoglobin, creatine kinase-MB (CK-MB), and troponin-I. It has also been used as an interfering agent to test the effectiveness of the Heparin Red kit in the detection of various heparins.

Biochem/physiol Actions

Bilirubin is an active oxidative DNA cleaving agent as well as an effective antioxidant agent, a hydroxyl radical quencher. This bile pigment has both antioxidant and toxic properties. It is a natural inhibitor of vascular smooth muscle cell proliferation (VSMCs). It displays anti-inflammatory and anti-proliferative properties when used as a therapeutic agent in lung/vascular diseases. Serum bilirubin concentration slightly above the normal levels have shown a lesser incidence of heart disease. It is a potential therapeutic agent in heart transplantation and T-cell mediated immune disorders.
Well over 99% of total bilirubin is transported as a conjugate with albumin. Abnormally high levels of bilirubin can cause severe neurological damage, but mildly elevated levels are linked to protection from oxidative stress.

Analysis Note

λmax. ∼453 nm, mol. absorption >56100 (chloroform)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenichiro Yamashita et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 18(6), 765-767 (2004-02-24)
Biliverdin, a product of heme oxygenase-1 (HO-1) enzymatic action, is converted into bilirubin, which has been considered a waste product in the past. We now show that administration of biliverdin has a salutary effect in organ transplantation. A brief course
Yen-Wen Wu et al.
Acta Cardiologica Sinica, 36(6), 649-659 (2020-11-26)
The rapid diagnosis of acute myocardial infarction (AMI) is a clinical and operational priority in emergency departments. Serial serum levels of cardiac biomarkers play a crucial role in the evaluation of patients presenting with acute chest pain, so that an
Stefan W Ryter et al.
American journal of respiratory cell and molecular biology, 36(2), 175-182 (2006-09-19)
Heme oxygenase (HO)-1, an inducible, low-molecular-weight stress protein, confers cellular and tissue protection in multiple models of injury and disease, including oxidative or inflammatory lung injury, ischemia/reperfusion (I/R) injuries, and vascular injury/disease. The tissue protection provided by HO-1 potentially relates
Robert Ollinger et al.
Cell cycle (Georgetown, Tex.), 6(1), 39-43 (2007-01-25)
We have recently shown that the natural bile pigment bilirubin has antiproliferative effects on vascular smooth muscle cells (VSMCs). Bilirubin is the end product of heme catabolism mediated by heme oxygenases and has for decades been considered a toxic waste
U Warttinger et al.
Analytical and bioanalytical chemistry, 408(28), 8241-8251 (2016-10-22)
Heparins are widely used anticoagulant drugs. The current monitoring practice for heparin in plasma, such as the chromogenic anti-factor Xa assay, relies on heparin-triggered activation of antithrombin, an inhibitor of coagulation proteases. Such assays are not applicable to the detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service