Skip to Content
Merck

137262

Tricine

EMPROVE® EVOLVE

Synonym(s):

Tricine, N-[Tris(hydroxymethyl)methyl]glycine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HOCH2)3CNHCH2CO2H
CAS Number:
Molecular Weight:
179.17
UNSPSC Code:
12161700
Beilstein/REAXYS Number:
1937804
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, heavy metals, trace metals, residual solvents, tested (
Endotoxin, microbial, and trace metals; tested)

pH

7.4-8.8

useful pH range

7.4-8.8

pKa (25 °C)

8.1

pKa 

8.1

suitability

suitable for manufacturing use

storage temp.

2-30°C

SMILES string

OCC(CO)(CO)NCC(O)=O

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

Application

Buffer component for separation of low molecular weight peptides.
Buffer component for separation of low molecular weight peptides.Tricine is a biological buffer often referred to as a “Good′s” buffer. The pKa of Tricine is 8.1 which makes Tricine is a good candidate for biological applications that require pH control above physiological. Tricine is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.Tricine can be used in a wide array of biological applications including as a component in media formulations. Specific applications include buffers for electrophoresis, protein purification and diagnostic reagent production.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Tricine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.