Skip to Content
Merck
All Photos(2)

Key Documents

10820

Sigma-Aldrich

Aprotinin from bovine lung

lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg

Synonym(s):

BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C284H432N84O79S7
CAS Number:
Molecular Weight:
6511.44
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

biological source

bovine lung

Assay

~80% (HPCE)

form

crystalline (fine)

quality

lyophilized

specific activity

≥3500 U/mg

mol wt

~6,500

technique(s)

electrophoresis: suitable

color

white

solubility

phosphate buffer: 5 mg/mL, clear, colorless (PH 7.6 (70 mm phosphate))

UniProt accession no.

storage temp.

2-8°C

SMILES string

S1SCC2NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C5NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C9N(CCC9)C(=O)CNC(=O)C(NC(=O)C(NC(=O)C%11N(CCC%11)C(=O

InChI

1S/C284H432N84O79S7/c1-21-144(9)222-271(439)337-174(68-46-105-309-282(300)301)239(407)340-187(120-160-77-85-164(374)86-78-160)251(419)341-185(116-156-55-29-24-30-56-156)250(418)342-188(121-161-79-87-165(375)88-80-161)252(420)346-191(123-208(291)378)246(41

InChI key

ZPNFWUPYTFPOJU-UHFFFAOYSA-N

Gene Information

cow ... PTI(404172)

Looking for similar products? Visit Product Comparison Guide

Application

Aprotinin is largely used as an inhibitor of trypsin.

Biochem/physiol Actions

Aprotinin is a competitive serine protease inhibitor that forms stable complexes with and blocks the active sites of enzyme. This binding is reversible, and most aprotinin-protease complexes will dissociate at extreme pH levels >10 or <3. Structurally, Aprotinin is a monomeric globular protein derived from bovine lung that consists of 58 amino acids, arranged in a single polypeptide chain with three crosslinking disulfide bridges.

Unit Definition

1 U corresponds to 1 biological kallikrein inhibitor unit (KIU)
One Trypsin Inhibitor Unit (TIU) will decrease the activity of two trypsin units by 50%, where one trypsin unit will hydrolyze 1.0 μmole of N-alpha-benzoyl-DL-arginine p-nitroanilide per minute at pH 7.8 and 25°C. Another commonly used unit is the KIU, with 1 TIU = 1,300 KIU.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Averna et al.
Methods in molecular biology (Clifton, N.J.), 1915, 195-206 (2019-01-09)
Immunoblotting is a procedure routinely used to analyze calpastatin expression. However, immunoblotting alone may not be adequate for this task, since calpastatin isoforms can vary by tissue, can be modified by partial digestion, and can undergo posttranslational modifications. Here we
Elizabeth J Osterlund et al.
Science advances, 8(16), eabm7375-eabm7375 (2022-04-21)
Cytoplasmic and membrane-bound BCL-2 family proteins regulate apoptosis, a form of programmed cell death, via dozens of binary protein interactions confounding measurement of the effects of inhibitors in live cells. In cancer, apoptosis is frequently dysregulated, and cell survival depends
Hamid Ebrahimi Orimi et al.
Scientific reports, 10(1), 9730-9730 (2020-06-18)
We introduced and validated a drop-on-demand method to print cells. The method uses low energy nanosecond laser (wavelength: 532 nm) pulses to generate a transient microbubble at the distal end of a glass microcapillary supplied with bio-ink. Microbubble expansion results in
Roberta De Tullio et al.
Methods in molecular biology (Clifton, N.J.), 1915, 163-185 (2019-01-09)
The production of recombinant calpastatin in E. coli has become an efficient tool to obtain discrete amounts of a specific calpastatin species that can be present concomitantly with other calpastatin fragments/forms in the same tissue or cell type in a
Robert E G Riddell et al.
Canadian journal of anaesthesia = Journal canadien d'anesthesie, 60(1), 16-23 (2012-11-08)
In light of the concerns about the safety of aprotinin, we wanted to determine if aprotinin use during cardiac surgery was associated with an increased risk of mortality and morbidity compared with the use of tranexamic acid (TXA). We hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service