Skip to Content
Merck

Skip To

09260

Atto 425-Streptavidin

BioReagent, suitable for fluorescence

Synonym(s):

Streptavidin-Atto 425

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352108
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥95% (HPCE)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

solubility

H2O: soluble

fluorescence

λex 437 nm; λem 477 nm in 0.1 M phosphate pH 7.0

suitability

corresponds for gel electrophoresis, suitable for fluorescence

storage temp.

−20°C

Application

Applied to the development of fluorescence binding assays to detect volatile organic compounds in both the liquid and gas phase[1].

Analysis Note

unconjugated dye ≤5% of total fluorescence

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
02744RSTREP-RO49937
suitability

corresponds for gel electrophoresis, suitable for fluorescence

suitability

-

suitability

-

suitability

corresponds for gel electrophoresis, suitable for fluorescence

fluorescence

λex 437 nm; λem 477 nm in 0.1 M phosphate pH 7.0

fluorescence

λex 655 nm; λem 680 nm in 0.1 M phosphate pH 7.0

fluorescence

-

fluorescence

-

solubility

H2O: soluble

solubility

DMF: soluble, DMSO: soluble, acetonitrile: soluble

solubility

water: 10 mg/L (mA405=100)

solubility

-

form

powder

form

powder

form

liquid

form

solid

assay

≥95% (HPCE)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters