Skip to Content
Merck

Skip To

09184

Dextran from Leuconostoc spp.

Mr ~100,000

Dextran from Leuconostoc spp.
Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
EC Number:
232-677-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Leuconostoc spp.)

Quality Segment

form

powder

mol wt

Mr ~100,000

mp

483 °C ((901 °F))

solubility

water: 1.5 g/10 mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

General description

May contain greater parts of lower molecular weight.

Application

Dextran is a branched glucan composed of linear α(1→6) linked glucose units and α (1→3) link initiated branches. Dextran ranges in size from 10,000 to 150,000 Kd. Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.

Other Notes

To gain a comprehensive understanding of our extensive range of dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
313923138731398
form

powder

form

powder

form

crystalline, solid

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

mol wt

Mr ~100,000

mol wt

Mr 450,000-650,000

mol wt

Mr 15,000-25,000

mol wt

Mr ~200,000

solubility

water: 1.5 g/10 mL, clear to slightly hazy, colorless to faintly yellow

solubility

water: 1.5 g/10 mL, clear, colorless

solubility

water: 0.5 g/5mL, colorless

solubility

-

biological source

(Leuconostoc spp.)

biological source

(Leuconostoc spp)

biological source

(Leuconostoc spp)

biological source

(leuconostoc mesenteroides)


Still not finding the right product?

Explore all of our products under Dextran from Leuconostoc spp.


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. It says on the discription "May contain greater parts of lower molecular weight." What is the molecular weight range?

    1 answer
    1. The bulk of this material has a molecular weight range of this material is 85 - 135 kDa. This information is lot specific and reported in the product Certificate of Analysis. A brief review of recent lots indicates 93 - 112 kDa molecular weights are typical. As with most polymers, there may be a portion of a powder sample that falls below the reported molecular weight value. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sigma/09184#product-documentation

      Helpful?

  2. What are the endotoxin specs for the dextran products?

    1 answer
    1. This product is not endotoxin tested. At this time there, is no Dextran product available that is endotoxin tested.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review