Skip to Content
Merck

05511

N-Acetyl-Arg-Gly-Lys(acetyl)-7-amido-4-methylcoumarin trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

N2-Acetyl-L-arginylglycyl-N3-acetyl-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-L-lysinamide trifluoroacetate salt, Ac-Arg-Gly-Lys(Ac)-AMC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H40N8O7 · xC2HF3O2
CAS Number:
Molecular Weight:
600.67 (free base basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32
Assay:
≥98% (HPLC)
Form:
powder or crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder or crystals

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(=O)NCCCC[C@H](NC(=O)CNC(=O)[C@@H](N)CCCNC(N)=N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C26H38N8O6.C2HF3O2/c1-15-12-23(37)40-21-13-17(8-9-18(15)21)33-25(39)20(7-3-4-10-30-16(2)35)34-22(36)14-32-24(38)19(27)6-5-11-31-26(28)29;3-2(4,5)1(6)7/h8-9,12-13,19-20H,3-7,10-11,14,27H2,1-2H3,(H,30,35)(H,32,38)(H,33,39)(H,34,36)(H4,28,29,31);(H,6,7)/t19-,20-;/m0./s1

InChI key

OOYFMABGIKUMHP-FKLPMGAJSA-N

Application

Novel fluorogenic assay for histone deacetylase activity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library