Skip to Content
Merck
All Photos(1)

Key Documents

04668

Sigma-Aldrich

O-Phospho-L-homoserine lithium salt

≥95% (TLC)

Synonym(s):

O-Phosphono-L-homoserine lithium salt, L-2-Amino-4-hydroxybutyric acid 4-phosphate lithium salt, L-Homoserine O-phosphate lithium salt, H-Hse(PO3H2)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10NO6P · xLi+
CAS Number:
Molecular Weight:
199.10 (free acid basis)
UNSPSC Code:
12352209
NACRES:
NA.26

Quality Level

Assay

≥95% (TLC)

form

powder

optical activity

[α]/D -18.0±2.0°, c = 1 in H2O

color

white to off-white

storage temp.

−20°C

SMILES string

[P](=O)(OCC[C@H](N)C(=O)O)(O)O

InChI

1S/C4H10NO6P/c5-3(4(6)7)1-2-11-12(8,9)10/h3H,1-2,5H2,(H,6,7)(H2,8,9,10)/t3-/m0/s1

InChI key

FXDNYOANAXWZHG-VKHMYHEASA-N

Biochem/physiol Actions

Metabolite in glycine, serine and threonine metabolism. In higher plants, O-phosphohomoserine represents a branch point between the methionine and threonine biosynthetic pathways.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Bartlem et al.
Plant physiology, 123(1), 101-110 (2000-05-12)
In higher plants, O-phosphohomoserine (OPH) represents a branch point between the methionine (Met) and threonine (Thr) biosynthetic pathways. It is believed that the enzymes Thr synthase (TS) and cystathionine gamma-synthase (CGS) actively compete for the OPH substrate for Thr and
An ATP-independent phosphotransferase in higher plants. The formation of O-phospho-L-homoserine from O-succinyl-L-homoserine and phosphate.
Haginiwa, J., et al.
Chemical & Pharmaceutical Bulletin, 27, 1710-1713 (1979)
O Fiehn et al.
Analytical chemistry, 72(15), 3573-3580 (2000-08-22)
Unknown compounds in polar fractions of Arabidopsis thaliana crude leaf extracts were identified on the basis of calculations of elemental compositions obtained from gas chromatography/low-resolution quadrupole mass spectrometric data. Plant metabolites were methoximated and silylated prior to analysis. All known
Oliver Kreft et al.
Plant physiology, 131(4), 1843-1854 (2003-04-15)
In plants, metabolic pathways leading to methionine (Met) and threonine diverge at the level of their common substrate, O-phosphohomoserine (OPHS). To investigate the regulation of this branch point, we engineered transgenic potato (Solanum tuberosum) plants affected in cystathionine gamma-synthase (CgS)
T Zhou et al.
Structure (London, England : 1993), 8(12), 1247-1257 (2001-02-24)
Homoserine kinase (HSK) catalyzes an important step in the threonine biosynthesis pathway. It belongs to a large yet unique class of small metabolite kinases, the GHMP kinase superfamily. Members in the GHMP superfamily participate in several essential metabolic pathways, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service