Skip to Content
Merck
All Photos(1)

Key Documents

04277

Sigma-Aldrich

Sodium phosphate tribasic dodecahydrate

≥98%

Synonym(s):

TSP, Trisodium phosphate (tert) dodecahydrate, Trisodium phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3PO4 · 12H2O
CAS Number:
Molecular Weight:
380.12
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98%

form

crystalline

impurities

≤0.001% heavy metals (as Pb)
≤2.5% free alkali (as NaOH)

solubility

boiling water: soluble 1 part
water: soluble 3.5 parts
alcohol: insoluble

density

1.62 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

As: ≤1 mg/kg
Fe: ≤10 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

InChI

1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3

InChI key

ASTWEMOBIXQPPV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate tribasic dodecahydrate was used to prepare buffer solution used in the analysis of pyronaridine in human whole blood and plasma by using high-performance liquid chromatography.

Biochem/physiol Actions

Sodium phosphate tribasic also referred to as Trisodium phosphate or TSP is a widely used cleaning agent. Trisodium phosphate disrupts cytoplasmic as well as outer membranes of Salmonella enterica cells and hence prevents the spread of bacteria. Further, it is corrosion inhibitor for iron and inhibits the rate of the anodic iron dissolution reaction in unbuffered solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behaviour of sodium silicate and sodium phosphate (tribasic) as corrosion inhibitors for iron.
R. D. Armstrong
J. Appl. Electrochem., 24, 1244-1248 (1994)
Y C Chen et al.
Journal of chromatography. B, Biomedical sciences and applications, 752(1), 39-46 (2001-03-20)
An improved high-performance liquid chromatography method using a diisopropyl-C14 reversed-phase column (Zorbax Bonus-RP column) and a liquid-liquid extraction technique with UV detection is presented for the analysis of pyronaridine in human whole blood and plasma. Tribasic phosphate buffer (50 mM
Balamurugan Sampathkumar et al.
Applied and environmental microbiology, 69(1), 122-129 (2003-01-07)
Trisodium phosphate (TSP) is now widely used during the processing of poultry and red meats, but the mechanism whereby it inactivates gram-negative bacteria such Salmonella spp. remains unclear. Thus, Salmonella enterica serovar Enteritidis (ATCC 4931) cells were treated with different
Santosh Lamichhane et al.
Analytical chemistry, 87(12), 5930-5937 (2015-05-20)
Metabolomic analyses of fecal material are gaining increasing attention because the gut microbial ecology and activity have an impact on the human phenotype and regulate host metabolism. Sample preparation is a crucial step, and in this study, we recommend a
Gisela Ferraro et al.
Journal of experimental botany, 66(11), 3381-3389 (2015-04-17)
Glutamate (Glu) is a taste enhancer that contributes to the characteristic flavour of foods. In fruit of tomato (Solanum lycopersicum L.), the Glu content increases dramatically during the ripening process, becoming the most abundant free amino acid when the fruit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service