Skip to Content
Merck
All Photos(1)

Key Documents

01480

Sigma-Aldrich

Acetylthiocholine iodide

≥99.0% (AT)

Synonym(s):

(2-Mercaptoethyl)trimethylammonium iodide acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COSCH2CH2N(CH3)3I
CAS Number:
Molecular Weight:
289.18
Beilstein:
3916578
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥99.0% (AT)

ign. residue

≤0.05%

mp

205-210 °C (lit.)
205-210 °C

solubility

H2O: 0.1 g/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[I-].CC(=O)SCC[N+](C)(C)C
[I-].CC(=O)SCC[N+](C)(C)C

InChI

1S/C7H16NOS.HI/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1

InChI key

NTBLZMAMTZXLBP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Acetylthiocholine iodide has been used as a substrate to determine anticholinesterase activity.
Substrate for the colorimetric determination of acetylcholinesterase activity.

Substrates

Acetylcholinesterase substrate and nicotinic acetylcholine receptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niko S Radulović et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 80, 114-129 (2015-03-15)
Herein we report on the comprehensive chemical analysis of the essential oils obtained from above- and underground parts of a previously unreported chemotype of Achillea falcata L. (Asteraceae) and, for the first time, on the biological/toxicological profile of its dominant/newly
Zrinka Kovarik et al.
Chemical research in toxicology, 28(5), 1036-1044 (2015-04-04)
Exposure to the nerve agent soman is difficult to treat due to the rapid dealkylation of the soman-acetylcholinesterase (AChE) conjugate known as aging. Oxime antidotes commonly used to reactivate organophosphate inhibited AChE are ineffective against soman, while the efficacy of
Belgin Sever et al.
International journal of biological macromolecules, 163, 1970-1988 (2020-09-16)
Alzheimer's disease (AD) is a complex, predominant, and progressive form of dementia. The treatment of AD alters depending on the cognitive and behavioral symptoms. The utility of cholinergic replacement by acetylcholinesterase (AChE) inhibitors in AD treatment has been well-documented so
J Blohberger et al.
Cell death & disease, 6, e1685-e1685 (2015-03-15)
Proliferation, differentiation and death of ovarian cells ensure orderly functioning of the female gonad during the reproductive phase, which ultimately ends with menopause in women. These processes are regulated by several mechanisms, including local signaling via neurotransmitters. Previous studies showed
Wei Li et al.
Fitoterapia, 98, 117-123 (2014-07-30)
Three new 2-(2-phenylethyl)chromone derivatives (1-3), together with thirteen known ones (4-16), were isolated from the EtOAc extract of Chinese agarwood induced by artificial holing, originating from Aquilaria sinensis (Lour.) Gilg (Thymelaeaceae). The chemical structures of the new compounds were identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service