Skip to Content
Merck
All Photos(3)

Key Documents

01409

Sigma-Aldrich

Lithium potassium acetyl phosphate

purum p.a., substrate (for phosphotransacetylase), ≥97.0% (NT)

Synonym(s):

Acetyl phosphate lithium potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOP(O)(OK)(OLi)
CAS Number:
Molecular Weight:
184.06
Beilstein:
5689718
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

grade

substrate (for phosphotransacetylase)
purum p.a.

Assay

≥97.0% (NT)

form

powder

mp

>300 °C (lit.)

solubility

water: 25 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li+].[K+].CC(=O)OP([O-])([O-])=O

InChI

1S/C2H5O5P.K.Li/c1-2(3)7-8(4,5)6;;/h1H3,(H2,4,5,6);;/q;2*+1/p-2

InChI key

RLQMPLKXFIXRCV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium potassium acetyl phosphate is an organometallic compound that is used as a phosphoryl group donor. It is commonly used in protein phosphorylation reactions.

Application

Lithium potassium acetyl phosphate has been used as a component of cocktail A in evaluating the activity of choline acetyltransferase in mice brain tissue homogenates.

Biochem/physiol Actions

Lithium potassium acetyl phosphate is suitable as a source of acetyl phosphate. It is suitable for reducing inorganic monophosphate by-product formation by replacing inorganic oligophosphates in phosphatase-catalyzed transphosphorylation.

Other Notes

Energy-rich compound for the regeneration of adenosine-5′-triphosphate, ATP, and other triphosphates

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic regeneration of adenosine 5'-triphosphate: acetyl phosphate, phosphoenolpyruvate, methoxycarbonyl phosphate, dihydroxyacetone phosphate, 5-phospho-alpha-D-ribosyl pyrophosphate, uridine-5'-diphosphoglucose.
D C Crans et al.
Methods in enzymology, 136, 263-280 (1987-01-01)
Sumonto Mitra et al.
Frontiers in aging neuroscience, 13, 714186-714186 (2021-09-04)
Gradual decline in cholinergic transmission and cognitive function occurs during normal aging, whereas pathological loss of cholinergic function is a hallmark of different types of dementia, including Alzheimer's disease (AD), Lewy body dementia (LBD), and Parkinson's disease dementia (PDD). Glial
Emiko Kinoshita-Kikuta et al.
PloS one, 10(7), e0132598-e0132598 (2015-07-08)
Hybrid sensor kinase, which contains a histidine kinase (HK) domain, a receiver domain, and a histidine-containing phosphotransmitter (HPt) domain, conveys signals to its cognate response regulator by means of a His-Asp-His-Asp phosphorelay. We examined the multistep phosphorelay of a recombinant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service