00541
Ruthenium Red
for microscopy, 85.0-115.0% (calc. on dry basis, AT)
Synonym(s):
Ammoniated ruthenium oxychloride
About This Item
Recommended Products
grade
for microscopy
Quality Level
Assay
85.0-115.0% (calc. on dry basis, AT)
form
powder
impurities
~5% water
solubility
water: 10 mg/mL, clear to turbid
εmax
≥150 at 529-539 nm
suitability
suitable for microscopy
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
N.N.N.N.N.N.N.N.N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++]O[Ru++]O[Ru++]
N.N.N.N.N.N.N.N.N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++]O[Ru++]O[Ru++]
InChI
1S/6ClH.14H3N.2O.3Ru/h6*1H;14*1H3;;;;;/q;;;;;;;;;;;;;;;;;;;;;;3*+2/p-6
InChI key
ZXYSDUPLHOJIOR-UHFFFAOYSA-H
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service