Skip to Content
Merck
All Photos(1)

Key Documents

00413

Sigma-Aldrich

DDMAB

≥94%

Synonym(s):

N-Dodecyl-N,N-(dimethylammonio)butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Quality Level

Assay

≥94%

mol wt

299.49 g/mol

CMC

4.3

SMILES string

CCCCCCCCCCCC[N+](C)(C)CCCC([O-])=O
CCCCCCCCCCCC[N+](C)(C)CCCC([O-])=O

InChI

1S/C18H37NO2/c1-4-5-6-7-8-9-10-11-12-13-16-19(2,3)17-14-15-18(20)21/h4-17H2,1-3H3

InChI key

ZFVNBVQBIZJUEY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dyeing acetate fibers with 1,4-diaminoanthraquinone using didodecyldimethylammonium bromide as the dye vehicle.
Ik-Soo, K.
Textile Res., 68(6), 422-427 (1998)
Talita L Santos et al.
Biophysical journal, 111(12), 2676-2688 (2016-12-22)
A large number of antimicrobial peptides (AMPs) acts with high selectivity and specificity through interactions with membrane lipid components. These peptides undergo complex conformational changes in solution; upon binding to an interface, one major conformation is stabilized. Here we describe
Tensioactive properties of zwitterionic carboxybetaine amphiphiles.
Chevalier, Y., et al.
Langmuir, 7, 848-853 (1991)
C Brenner et al.
Analytical biochemistry, 224(2), 515-523 (1995-01-20)
The ability to extract mycoplasma membrane protein antigens using the alkyl carboxybetaine surfactants (N-dodecyl-N,N-dimethylammonio)butyrate (DDMAB, CMC = 4.3 mM) and (N-dodecyl-N,N-dimethylammonio)undecanoate (DDMAU, CMC = 0.13 mM) was assessed by protein titration and SDS-PAGE analysis. The maximum yields of membrane protein
S Welling-Wester et al.
Journal of chromatography. A, 816(1), 29-37 (1998-09-19)
Detergents (surfactants) are the key reagents in the extraction and purification of integral membrane proteins. Zwitterionic and non-ionic detergents were used for the extraction of recombinant glycoprotein D (gD-1) of herpes simplex virus type 1 (HSV-1) from insect cells infected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service