Skip to Content
Merck
All Photos(4)

Key Documents

S1878

Sigma-Aldrich

Sodium (meta)periodate

≥99.0%

Synonym(s):

Sodium periodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder
solubility:
H2O: 50 mg/mL

grade

for analytical purposes

Assay

≥99.0%

form

powder

reaction suitability

reagent type: oxidant

pH

3.5-5.5 (25 °C, 107 g/L)

solubility

H2O: 50 mg/mL

SMILES string

[Na+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium (meta)periodate is commonly used as an oxidizing agent in organic synthesis for the oxidation of various organic substrates. It can also be used as a co-oxidant with other oxidizing reagents such as osmium tetroxide and potassium permanganate. The oxidation of sodium periodate takes place in a water medium, but an organic co-solvent is used for organic substrates that are insoluble in water.

Application

Sodium (meta)periodate can be used as an oxidizing agent for the oxidation of:      
  • 1,2-diols to aldehydes or ketones via oxidative cleavage.       Sulfides to sulfoxides in the presence of phase transfer catalysts.      
  • Diaryl, dialkyl, and aryl alkyl selenides to the respective selenoxides.      
  • Phenols and their derivatives to corresponding quinones.
  • Acylphosphoranes to α,β-dicarbonyl compounds.

It can also be used in the:      
  • Selective alkene epoxidation and alkane hydroxylation,·
  • Oxidation of carbohydrates.      
  • Iodination of aromatic compounds.

Useful reagent for the oxidation of carbohydrates prior to labeling with biotin hydrazide

Features and Benefits

NaIO4 is a stable, selective, and cost-effective reagent that is capable of reacting under mild conditions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jusung Oh et al.
Theranostics, 9(14), 4182-4191 (2019-07-10)
Cerebrospinal fluid (CSF) leakage can lead to brain and spine pathologies and there is an urgent need for a rapid diagnostic method for determining CSF leakage. Beta-2 transferrin (β2TF), asialotransferrin, is a specific CSF glycoprotein biomarker used to determine CSF
Zhilong Chen et al.
Molecular therapy. Methods & clinical development, 17, 21-33 (2020-01-01)
The motor system controls muscle movement through lower motor neurons in the spinal cord and brainstem. Lower motor neurons are efferent neurons in the central nervous system (CNS) characterized by axonal projections that reach specific targets in the periphery. Lower
Wei-Yao Chen et al.
Frontiers in endocrinology, 12, 572690-572690 (2021-03-30)
The global obesity population is increasing year-by-year, and the related cost is sharply increasing annually. There are several methods available to combat obesity; however, there is a lack of a single tool that is both safe and efficacious. The use
Zhao Wei et al.
Cell stem cell, 27(5), 798-812 (2020-09-16)
Matrix dynamics influence how individual cells develop into complex multicellular tissues. Here, we develop hydrogels with identical polymer components but different crosslinking capacities to enable the investigation of mechanisms underlying vascular morphogenesis. We show that dynamic (D) hydrogels increase the
Maria Molinos et al.
Biomacromolecules, 13(2), 517-527 (2012-02-01)
Dextrin, a glucose polymer with low molecular weight, was used to develop a fully resorbable hydrogel, without using chemical initiators. Dextrin was first oxidized (oDex) with sodium periodate and then cross-linked with adipic acid dihidrazide, a nontoxic cross-linking molecule. Furthermore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service