Skip to Content
Merck
All Photos(3)

Key Documents

S0505

Sigma-Aldrich

Sodium sulfite

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥98%
form:
powder or crystals
Pricing and availability is not currently available.

Agency

suitable for SM 5210

Quality Level

Assay

≥98%

form

powder or crystals

pH

9.0-10.5 (25 °C, 126 g/L)

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfite is a water-soluble inorganic salt that is commonly used as a reagent for sulfonation and sulfomethylation reactions. Additionally, it is also used as a bleaching, de-sulfurizing, and dechlorinating agent.

Application

Sodium sulfite can be used as:      
  • A source of sulfur dioxide (SO2) in the synthesis of 1,4 diazabicyclo[2.2.2]octane bis(sulfur dioxide) (DABSO).      
  • A reducing agent in the photocatalytic cyclization of 2-isocyanoaryl thioethers to synthesize 2-CF2/CF3-containing benzothiazoles in the presence of Ir-based catalyst.     
  • A reagent in the selective synthesis of 2-amino-1-iodonaphthalene from 2-naphthylamine in the presence of NaIO3/HCl.

In combination with carboxylic acids, it forms an effective system for the reductive acylation of aromatic nitro compounds.

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insu Lee et al.
Micromachines, 10(1) (2019-01-02)
Hypoxic environment is known as one of the critical factors in various physiological/pathological processes. It is imperative to recapitulate oxygen level in microscale for human physiology/pathology induced by hypoxia. Herein, we propose an oxygen-regulating system that can be applied to
Gun-Soo Park et al.
The Journal of molecular diagnostics : JMD, 22(6), 729-735 (2020-04-11)
The coronavirus disease 2019 (COVID-19) pandemic now has >2,000,000 confirmed cases worldwide. COVID-19 is currently diagnosed using quantitative RT-PCR methods, but the capacity of quantitative RT-PCR methods is limited by their requirement of high-level facilities and instruments. We developed and
Yoan Arribat et al.
Frontiers in cell and developmental biology, 7, 282-282 (2019-12-12)
Organogenesis is well characterized in vertebrates. However, the anatomical and functional development of intracellular compartments during this phase of development remains unknown. Taking an organellogenesis point of view, we characterize the spatiotemporal adaptations of the mitochondrial network during zebrafish embryogenesis.
Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Marcel E Sayre et al.
The Journal of comparative neurology, 527(14), 2371-2387 (2019-03-13)
Paired centers in the forebrain of insects, called the mushroom bodies, have become the most investigated brain region of any invertebrate due to novel genetic strategies that relate unique morphological attributes of these centers to their functional roles in learning

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. 해당 sodium sulfite 제품의 평균 입자 사이즈를 알고싶습니다. 답변 부탁드립니다.

    1 answer
    1. The average particle size for this product has not been determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service