Skip to Content
Merck
All Photos(3)

Documents

P5271

Sigma-Aldrich

Potassium dichromate

ReagentPlus®, ≥99.5%

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

product line

ReagentPlus®

Assay

≥99.5%

form

powder

reaction suitability

reagent type: oxidant

pH

3.6 (100 g/L)

mp

398 °C (lit.)

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium dichromate can be used as an oxidant:
  • For the oxidation of alcohols to aldehydes.
  • In the graft-polymerization of polyacrylonitrile film with polyaniline.
  • To synthesize oxidized nanocellulose.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface modification of polyacrylonitrile film by anchoring conductive polyaniline and determination of uricase adsorption capacity and activity
Bayramouglu G, et al.
Applied Surface Science, 256, 6710-6716 (2010)
Kyra T Newmaster et al.
Nature communications, 11(1), 1885-1885 (2020-04-22)
The oxytocin receptor (OTR) plays critical roles in social behavior development. Despite its significance, brain-wide quantitative understanding of OTR expression remains limited in postnatally developing brains. Here, we develop postnatal 3D template brains to register whole brain images with cellular
Sofia Abassi et al.
Environmental toxicology, 34(9), 1013-1024 (2019-05-17)
The freshwater green algae Closterium ehrenbergii has been considered as a model for eco-toxicological assessment in aquatic systems. Heat shock proteins (HSPs) are a class of highly conserved proteins produced in all living organisms, which participate in environmental stress responses.
Katlijn Vints et al.
Scientific reports, 9(1), 130-130 (2019-01-16)
Analysis of neuronal arborization and connections is a powerful tool in fundamental and clinical neuroscience. Changes in neuronal morphology are central to brain development and plasticity and are associated with numerous diseases. Golgi staining is a classical technique based on
Fu-Jun Liu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 110(2), 307-318 (2009-06-03)
Werner syndrome is a premature aging disorder characterized by cancer predisposition that is caused by loss of the Werner syndrome protein (WRN) helicase/exonuclease DNA repair protein. Hexavalent chromium is an environmental carcinogen and genotoxicant that is associated with respiratory cancers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service