Skip to Content
Merck
All Photos(2)

Key Documents

O0136

Sigma-Aldrich

Sodium oxalate

BioXtra

Synonym(s):

Ethanedioic acid sodium salt, Oxalic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

product line

BioXtra

Assay

≥99.0% (titration)

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.002%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluoroalkane aromatization over hot sodium oxalate.
McAlexander LH, et al.
Journal of Fluorine Chemistry, 99(1), 67-72 (1999)
Photoelectrocatalytic degradation of sodium oxalate by TiO2/Ti thin film electrode.
Chang CY, et al.
International Journal of Photoenergy, 2012 (2012)
Sodium oxalate structure refinement.
Reed DA and Olmstead MM.
Acta Crystallographica Section B, Structural Science, 37(4), 938-939 (1981)
Hong-Joo Lee et al.
Bioresource technology, 130, 211-217 (2013-01-12)
The present study investigated the feasibility of the recovery and reuse oxalic acid in a multistage process for the pretreatment of a lignocellulosic biomass. Electrodialysis (ED), an electrochemical process using ion exchange membranes, was used to recover and reuse oxalic
Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service