Skip to Content
Merck
All Photos(1)

Key Documents

71883

Sigma-Aldrich

Sodium peroxide

for Wurzschmitt-decomposition, ACS reagent, beads (small), ≥95%

Synonym(s):

Sodium superoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥95% (RT)
≥95%
grade:
ACS reagent
for Wurzschmitt-decomposition
form:
beads (small)

grade

ACS reagent
for Wurzschmitt-decomposition

Quality Level

Assay

≥95% (RT)
≥95%

form

beads (small)

impurities

≤0.003% total nitrogen (N)

pH

12.8 (20 °C, 100 g/L)

mp

460 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤20 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium peroxide (Sodium superoxide) has been synthesized by reacting sodium atoms and oxygen molecules at high dilution in argon. Its IR spectral studies have been reported. It is a strong oxidizing agent.

Application

Sodium peroxide may be used for the isotope dilution inductively coupled plasma mass spectrometric quantification of Ru, Pd, Ir and Pt in various geological samples. It may be employed for the inductively coupled plasma mass spectrometric quantification of rare earth elements (such as Y, Th, Zr, Hf, Nb and Ta) in geological reference materials.

Analysis Note

may contain particles (Fe, Ni)

Other Notes

Wurzschmitt decomposition; decomposition of ores by fusion

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Eagleson M.
Concise Encyclopedia Chemistry, 1080-1080 (1994)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Determination of platinum, palladium, ruthenium and iridium in geological samples by isotope dilution inductively coupled plasma mass spectrometry using a sodium peroxide fusion and tellurium coprecipitation
Enzweiler J, et al.
Analyst, 120(5), 1391-1396 (1995)
Infrared spectra and bonding in the sodium superoxide and sodium peroxide molecules.
Andrews L.
The Journal of Physical Chemistry, 73(11), 3922-3928 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service