Skip to Content
Merck
All Photos(1)

Key Documents

69850

Sigma-Aldrich

Molybdenum(VI) oxide

puriss. p.a., 99.5%

Synonym(s):

Molybdenum trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoO3
CAS Number:
Molecular Weight:
143.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.5%
form:
powder

grade

puriss. p.a.

Quality Level

Assay

99.5%

form

powder

mp

795 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤10 mg/kg
Co: ≤20 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤10 mg/kg
NH4+: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤20 mg/kg
Pb: ≤20 mg/kg
Zn: ≤20 mg/kg

SMILES string

O=[Mo](=O)=O

InChI

1S/Mo.3O

InChI key

JKQOBWVOAYFWKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molybdenum(VI) oxide (MoO3) is an acidic metal(VI) oxide. It is widely employed in selective oxidation reactions. Its temperature-programmed reduction has been reported. Kinetics and mechanism of its reduction in the presence of hydrogen, propene, butene-1 and CO has been investigated. It is reduced to metal upon reaction with hydrogen at high temperatures.

Application

Molybdenum(VI) oxide (MoO3) has been used as a starting material for the deposition of Mo (Molybdenum) over the MgO-330 and MgO-500 supports by employing methanol as solvent.
Precursor to LAMOX fast ion conductors and superconductors.
Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13 and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of molybdenum (VI) oxide reduction.
Sloczynski J.
Journal of Solid State Chemistry, 118(1), 84-92 (1995)
Eagleson M.
Concise Encyclopedia Chemistry, 509-509 (1994)
Temperature-programed reduction of molybdenum (VI) oxide and molybdenum (IV) oxide.
Arnoldy P, et al.
The Journal of Physical Chemistry, 89(21), 4517-4526 (1985)
Eagleson M.
Concise Encyclopedia Chemistry, 224-224 (1994)
High-activity MgO-supported CoMo hydrodesulfurization catalysts prepared by non-aqueous impregnation.
Kaluza L, et al.
Applied Catalysis. B, Environmental, 162, 430-436 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service