Skip to Content
Merck
All Photos(1)

Key Documents

676845

Sigma-Aldrich

Diethyl ether

≥98.0%, ACS reagent, contains ≤2% ethanol and ≤10ppm BHT as inhibitor

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.07
grade:
ACS reagent
bp:
34.6 °C (lit.)
vapor pressure:
28.5 psi ( 55 °C)
8.38 psi ( 20 °C)

Product Name

Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor

grade

ACS reagent

Quality Level

vapor density

2.6 (vs air)

vapor pressure

28.5 psi ( 55 °C)
8.38 psi ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

320 °F

contains

≤2% ethanol and ≤10ppm BHT as inhibitor

expl. lim.

36.5 %

dilution

(for analytical testing)

impurities

≤0.0002 meq/g Titr. acid
≤0.001% Carbonyl (as HCHO)
≤0.5% (water)
≤1 ppm Peroxide (as H2O2)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl ether (also known as ethyl ether or ether) is a laboratory solvent. It is a desirable solvent for oils, fats, waxes, perfumes, dyes, resins, hydrocarbons, and gums. It is also used as fuel and to synthesize other chemical compounds. DEE is widely employed as a solvent for Grignard synthesis.

Application

Diethyl ether is used as a solvent in the 3D-printed reaction ware device for chemical synthesis.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inês Mesquita et al.
Cell reports, 30(12), 4052-4064 (2020-03-27)
Hypoxia-inducible factor-1 alpha (HIF-1α) is considered a global regulator of cellular metabolism and innate immune cell functions. Intracellular pathogens such as Leishmania have been reported to manipulate host cell metabolism. Herein, we demonstrate that myeloid cells from myeloid-restricted HIF-1α-deficient mice
Alessandra Forni et al.
Journal of molecular graphics & modelling, 38, 31-39 (2012-10-23)
The solvent effect on the I⋯O halogen bonding in complexes of iodobenzene derivatives with formaldehyde has been investigated by systematically varying the substituents on the iodobenzene ring. Calculations have been performed at MP2 and DFT levels of theory, using the
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Faysal Benaskar et al.
ChemSusChem, 6(2), 353-366 (2012-11-30)
A μ(2)-process in the Ullmann-type C-O coupling of potassium phenolate and 4-chloropyridine was successfully performed in a combined microwave (MW) and microflow process. Selective MW absorption in a micro-fixed-bed reactor (μ-FBR) by using a supported Cu nanocatalyst resulted in an
Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service