Skip to Content
Merck
All Photos(1)

Key Documents

63537

Sigma-Aldrich

Manganese(II) acetate tetrahydrate

purum p.a., ≥99.0% (KT)

Synonym(s):

Manganous acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mn · 4H2O
CAS Number:
Molecular Weight:
245.09
Beilstein:
3730728
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (KT)
form:
crystals

vapor pressure

<1 hPa ( 25 °C)

Quality Level

grade

purum p.a.

Assay

≥99.0% (KT)

form

crystals

pH

7 (20 °C, 50 g/L)

mp

>300 °C (lit.)

density

1.589 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Mn]OC(C)=O

InChI

1S/2C2H4O2.Mn.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

CESXSDZNZGSWSP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II) acetate tetrahydrate is a hydrated salt of manganese. Kinetic studies suggest that its dehydration takes place in two-step reaction involving the loss of two water molecules. Its crystal structure has been investigated by X-ray studies. Its crystals exhibit monoclinic crystal system and space group P21/c.

Application

Manganese(II) acetate tetrahydrate may be used in the sol gel synthesis of doped TiO2 containing different amounts of Mn2+.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of manganese acetate tetrahydrate.
Bertaut EF, et al.
Acta Crystallographica Section B, Structural Crystallography and Crystal Chemistry, 30(9), 2234-2236 (1974)
Kinetic analysis of thermal decomposition reactions: Part VI. Thermal decomposition of manganese (II) acetate tetrahydrate.
Diefallah EHM.
Thermochimica Acta, 202, 1-16 (1992)
Influence of manganese ions on the anatase-rutile phase transition of TiO2 prepared by the sol-gel process.
Arroyo R, et al.
Materials Letters, 54(5), 397-402 (2002)
Kyu-Nam Jung et al.
Scientific reports, 5, 7665-7665 (2015-01-08)
Rechargeable metal-air batteries are considered a promising energy storage solution owing to their high theoretical energy density. The major obstacles to realising this technology include the slow kinetics of oxygen reduction and evolution on the cathode (air electrode) upon battery
Qiangqiang Sun et al.
Journal of hazardous materials, 286, 276-284 (2015-01-16)
Comparative experiments were conducted to investigate the catalytic ability of MnO(x)/SBA-15 for the ozonation of clofibric acid (CA) and its reaction mechanism. Compared with ozonation alone, the degradation of CA was barely enhanced, while the removal of TOC was significantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service