Skip to Content
Merck
All Photos(1)

Key Documents

496219

Sigma-Aldrich

N,N-Diisopropylethylamine

99.5%, biotech. grade

Synonym(s):

N-Ethyldiisopropylamine, DIPEA, Ethyldiisopropylamine, ‘Hünig’s base’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NC2H5
CAS Number:
Molecular Weight:
129.24
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.21
grade:
biotech. grade
Assay:
99.5%
application(s):
peptide synthesis
bp:
127 °C (lit.)
vapor pressure:
31 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

grade

biotech. grade

Quality Level

vapor pressure

31 mmHg ( 37.7 °C)

Assay

99.5%

form

liquid

impurities

<0.050% water

color

APHA: <20

refractive index

n20/D 1.414 (lit.)

pH

12.3 (20 °C)

bp

127 °C (lit.)

mp

<−50 °C (lit.)

solubility

water: soluble 4.01 g/L at 20 °C

density

0.742 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCN(C(C)C)C(C)C

InChI

1S/C8H19N/c1-6-9(7(2)3)8(4)5/h7-8H,6H2,1-5H3

InChI key

JGFZNNIVVJXRND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N, N-Diisopropylethylamine is a hindered non-nucleophilic amine base used in alkylation, aldol-like reactions, eliminations, and selective generation of enolates. It is widely used as a proton scavenger in organic synthesis. Due to its less nucleophilic nature, it produces less byproducts.
N, N-Diisopropylethylamine is a hindered non-nucleophilic amine base used in alkylation, aldol-like reactions, eliminations, and selective generation of enolates. It is widely used as a proton scavenger in organic synthesis. Due to its less nucleophilic nature, it produces less byproducts.

Application

N, N-Diisopropylethylamine can be used as a base:
  • In the palladium (0)-catalyzed alkoxy carbonylation of allyl phosphates and acetate.
  • In the selective enolate formation along with boryl triflates.
  • In the C–N coupling of aryl halides with nitroarenes in the presence of nickel catalyst.

N,N-Diisopropylethylamine may be used in the synthesis of mannosylated ovalbumin peptides.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.1 °F

Flash Point(C)

9.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Hydrogenation of α,β-Unsaturated Nitriles with Base-Activated Iridium N, P Ligand Complexes
Muller MA and Pfaltz A.
Angewandte Chemie (International Edition in English), 126(33), 8812-8815 (2014)
Facile and Gram-scale Synthesis of Metal-free Catalysts: Toward Realistic Applications for Fuel Cells.
Kim OH, et al.
Scientific Reports, 5 (2015)
Lipase-catalysed synthesis of olvanil in organic solvents.
Reyes-Duarte D, et al.
Biotechnology Letters, 24(24), 2057-2061 (2002)
Alexandre Yersin et al.
Biophysical journal, 94(1), 230-240 (2007-09-18)
Interaction between the iron transporter protein transferrin (Tf) and its receptor at the cell surface is fundamental for most living organisms. Tf receptor (TfR) binds iron-loaded Tf (holo-Tf) and transports it to endosomes, where acidic pH favors iron release. Iron-free
Irina Naletova et al.
Cancers, 11(9) (2019-09-11)
Angiogenin (ANG), an endogenous protein that plays a key role in cell growth and survival, has been scrutinised here as promising nanomedicine tool for the modulation of pro-/anti-angiogenic processes in brain cancer therapy. Specifically, peptide fragments from the putative cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service