Skip to Content
Merck
All Photos(1)

Key Documents

417661

Sigma-Aldrich

Potassium hydroxide solution

45 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
liquid

grade

for analytical purposes

form

liquid

concentration

43.0-47.0 wt. % (KOH basis, by HCl, titration)
45 wt. % in H2O

density

1.456 g/mL at 25 °C

SMILES string

[K+].[O-][H]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide (KOH) solution is a strong alkali widely used in chemical synthesis. Van Slyke apparatus and Hildebrand adsorption technique have been utilized to determine the solubility of oxygen in different concentration of KOH solution.

Application

Potassium hydroxide solution may be used in the following processes:
  • Depolymerization of poly(ethylene terephthalate) (PET) flakes.
  • Synthesis of potassium bicarbonate and potassium carbonate.
  • As an eluent in the nonsuppressor ion chromatography to separate and determine inorganic and organic anions.
  • Saponification of fruit seed oil to determine tocopherols and tocotrienols.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nonsuppressor ion chromatography of inorganic and organic anions with potassium hydroxide as eluent.
Okada T and Kuwamoto T.
Analytical Chemistry, 55(7), 1001-1004 (1983)
The solubility and diffusion coefficient of oxygen in potassium hydroxide solutions.
Davis RE, et al.
Electrochimica Acta, 12(3), 287-297 (1967)
Native oils from apple, blackcurrant, raspberry, and strawberry seeds as a source of polyenoic fatty acids, tocochromanols, and phytosterols: a health implication.
Pieszka M, et al.
Journal of Chemistry, 2015 (2015)
Kinetics of depolymerization of poly (ethylene terephthalate) in a potassium hydroxide solution.
Wan BZ, et al.
Industrial & Engineering Chemistry Research, 40(2), 509-514 (2001)
Ledgard J.
The Preparatory Manual of Explosives., 79-79 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service