Skip to Content
Merck
All Photos(1)

Key Documents

411515

Sigma-Aldrich

Lithium bromide solution

54 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBr
CAS Number:
Molecular Weight:
86.85
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

form

liquid

concentration

49.0-59.0% (silver nitrate titration)
54 wt. % in H2O

density

1.57 g/mL at 25 °C

SMILES string

[Li+].[Br-]

InChI

1S/BrH.Li/h1H;/q;+1/p-1

InChI key

AMXOYNBUYSYVKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium bromide solution contains 54 wt.% lithium bromide in water. The vapor pressure of the aqueous solution of lithium bromide has been studied over a range of temperature. Lithium bromide solution forms the absorber in the absorption refrigerator. Its pool boiling heat transfer coefficient has been obtained. It is a corrosive medium on metals.

Application

Lithium bromide solution may be used to dissolve/regenerate cellulose to form cellulose gel. It may also be used as a solvent for corrosion studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic properties of aqueous electrolyte solutions. 1. Vapor pressure of aqueous solutions of lithium chloride, lithium bromide, and lithium iodide.
Patil KR, et al.
Journal of Chemical and Engineering Data, 35(2), 166-168 (1990)
Use of differential pulse polarography to study corrosion of galvanized steel in aqueous lithium bromide solution.
Garcia-Anton J, et al.
Corrosion (Houston, TX, United States), 50(2), 91-97 (1994)
Corrosion behavior and galvanic studies of brass and bronzes in aqueous lithium bromide solutions.
Mu?oz AI, et al.
Corrosion (Houston, TX, United States), 58(7), 560-569 (2002)
Cellulose dissolution in aqueous lithium bromide solutions.
Yang YJ, et al.
Cellulose, 21(3), 1175-1181 (2014)
Pool boiling performance of lithium bromide solution on enhanced tubes.
Sim YS and Kim NH.
Journal of Mechanical Science and Technology, 29(6), 2555-2563 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service