Skip to Content
Merck
All Photos(1)

Key Documents

407666

Sigma-Aldrich

Ammonia solution

0.4 M in dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
Pricing and availability is not currently available.

vapor pressure

11.73 psi ( 55 °C)
4.16 psi ( 20 °C)

Quality Level

form

liquid

concentration

0.4 M in dioxane
450-600 mmol/L (titration)

density

1.023 g/mL at 25 °C

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a 0.4 M solution of ammonia in dioxane. Ammonia is a colorless gas with a characteristic strong odor. It is highly soluble in water, alcohol, chloroform and ether. It is extensively sold in the form of solution.1 Ammonia molecule is pyramidal in shape with nitrogen in the center and the three hydrogen atoms along the three vertices and the lone pair of electrons on the fourth vertex. Ammonia solution is commonly used as a base.[1]

Application

Ammonia solution (0.4 M in dioxane) may be used in the synthesis of 8-nitroquinolin-2-amine[2] and silphenylene-containing polymers.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a new antileishmanial hit in 8-nitroquinoline series.
Paloque L, et al.
European Journal of Medicinal Chemistry, 54, 75-86 (2012)
Eagleson M.
Concise Encyclopedia Chemistry, 65-66 (1994)
Synthesis and properties of polymers containing silphenylene moiety via catalytic cross-dehydrocoupling polymerization of 1,4-bis (dimethylsilyl) benzene.
Li Y and Kawakami Y.
Macromolecules, 32(26), 8768-8773 (1999)
Patrick M Erwin et al.
The ISME journal, 8(3), 575-588 (2013-10-25)
Ascidians are ecologically important components of marine ecosystems yet the ascidian microbiota remains largely unexplored beyond a few model species. We used 16S rRNA gene tag pyrosequencing to provide a comprehensive characterization of microbial symbionts in the tunic of 42
S M Gråsten et al.
The Journal of nutrition, 130(9), 2215-2221 (2000-08-26)
Cereal fiber may reduce the risk of colorectal cancer by diluting colonic contents due to increased fecal output, by accelerating intestinal transit, by increasing fecal frequency and by altering bacterial metabolism. The effects of whole-meal rye bread on some putative

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service