Skip to Content
Merck
All Photos(2)

Key Documents

402966

Sigma-Aldrich

Lithium metaborate

ACS reagent, ≥98.0%

Synonym(s):

Boric acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBO2
CAS Number:
Molecular Weight:
49.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

≥98.0%
98.0-102.0% (ACS specification)

form

powder

impurities

≤0.004% P compounds
≤0.01% insolubles

loss

≤2.0% loss on fusion, 950°C

mp

845 °C (lit.)

bulk density

≥0.25 g/mL

cation traces

Al: ≤0.001%
Ca: ≤0.01%
Fe: ≤0.002%
K: ≤0.005%
Mg: ≤5 ppm
Na: ≤0.005%
Si: ≤0.01%
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[Li+].[O-]B=O

InChI

1S/BO2.Li/c2-1-3;/q-1;+1

InChI key

HZRMTWQRDMYLNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium metaborate is an alkali borate. Crystal structure study reveals it has monoclinic structure with space group P21/c. It has been synthesized using orthoboric acid and lithium carbonate. Its vapor pressure has been analyzed using torsion-effusion method.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 24 trace elements in felsic rocks by inductively coupled plasma mass spectrometry after lithium metaborate fusion.
Panteeva SV, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 58(2), 341-350 (2003)
Thermodynamics of Vaporization of Lithium Oxide, Boric Oxide, and Lithium Metaborate.
Hildenbrand DL, et al.
J. Chem. Phys. , 39(2), 296-301 (1963)
Pressure-induced coordination changes in LiBO2.
Lei L, et al.
Journal of Solid State Chemistry, 182(11), 3041-3048 (2009)
The crystal structure of lithium metaborate.
Zachariasen WH.
Acta Crystallographica, 17(6), 749-751 (1964)
S Miljanić et al.
Radiation protection dosimetry, 119(1-4), 191-196 (2006-05-20)
Recently, two new types of 'tissue equivalent' thermoluminescent detectors (TLDs) have aroused attention: LiF:Mg,Cu,Na,Si and Li2B4O7:Cu,Ag,P. In this work the characteristics of both detectors were compared with the characteristics of the well-known type LiF:Mg,Ti detector, TLD-100. The following properties were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service