Skip to Content
Merck
All Photos(1)

Key Documents

38270

Sigma-Aldrich

Diisopropyl ether

puriss. p.a., ≥98.5% (GC)

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.07
Assay:
≥98.5% (GC)
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)

vapor density

3.5 (vs air)

Quality Level

vapor pressure

120 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

827 °F

contains

~0.001% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

1-21 %, 100 °F

impurities

≤0.001% peroxides (as H2O2)
≤0.005% free acid (as C2H5COOH)
≤0.1% water
≤0.5% isopropanol (GC)

evapn. residue

≤0.002%

refractive index

n20/D 1.367 (lit.)
n20/D 1.368

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.724 g/mL at 20 °C
0.725 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisopropyl ether is a colorless liquid. It is widely used as a solvent for the isolation of fats and oils. It can be synthesized from isopropyl alcohol. Its mixture with isooctane has been added to the gasoline fuel as an antiknocking agent. Its synthesis from propylene and water in a single-pass trickle-bed reactor in the presence of an ion-exchange resin catalyst Amberlyst 15 has been proposed.

Application

Diisopropyl ether may be employed as a solvent for the preparation of aliphatic and aromatic cyanohydrins.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 325-325 (1994)
Single stage synthesis of diisopropyl ether-an alternative octane enhancer for lead-free petrol.
Heese FP, et al.
Catalysis Today, 49(1), 327-335 (1999)
Novel (R)-oxynitrilase sources for the synthesis of (R)-cyanohydrins in diisopropyl ether.
Kiljunen E and Kanerva LT.
Tetrahedron Asymmetry, 8(8), 1225-1234 (1997)
Fiorella Belpoggi et al.
Annals of the New York Academy of Sciences, 982, 70-86 (2003-02-04)
Tert-amyl-methyl ether (TAME) was administered by gavage in extra virgin olive oil solution at concentrations of 750, 250, or 0 mg/kg bw to groups of 100 male and 100 female Sprague-Dawley rats 8 weeks old at the start of the
I Linhart et al.
Toxicology and applied pharmacology, 136(1), 155-160 (1996-01-01)
Biotransformation of acrolein (ACR) was studied in vivo in the rat following inhalation and ip administration. The major and minor urinary metabolites were 3-hydroxypropylmercapturic acid (HPMA) and 2-carboxyethylmercapturic acid (CEMA), respectively. Male Wistar rats were exposed to ACR, 23, 42

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service