Skip to Content
Merck
All Photos(3)

Documents

360589

Sigma-Aldrich

Tetrahydrofuran

≥99.0%, ACS reagent, contains 250 ppm BHT as inhibitor, suitable for HPLC

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.21

product name

Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%

grade

ACS reagent

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as inhibitor

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable

impurities

≤0.015% peroxide (as H2O2)
≤0.05% water

evapn. residue

≤0.03%

color

APHA: ≤20

refractive index

n20/D 1.407 (lit.)

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

solubility

water: soluble

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is a heterocyclic compound (cyclic ether). It is colorless, has low viscosity, and good solubility in a wide range of solvents. It is widely used as a solvent in organic synthesis, being very popular in reactions with organometallic compounds and Grignard reagents. Due to organic peroxides formation on long term storage, THF is usually stabilized by adding butylated hydroxytoluene (BHT). BHT removes the free radicals required for the peroxide formation.

Application

Tetrahydrofuran (THF) is used as a solvent in the following processes:

  • Organic synthesis
a) Grignard
b) Organometallic compounds
c) Reformatsky
d) Lithiation
e) Hydride reduction
f) Metal-catalyzed coupling (Heck, Stile, Suzuki)
g) Lewis acid mediated reactions
  • Crystallization
  • Polymerization. Ex. RAFT (Reversible Addition-Fragmentation Chain Transfer) polymerization of p-acetoxystyrene
  • Coatings
  • As an O-donor ligand to form coordination complexes
  • As mobile phase solvent in high-performance liquid chromatography

Features and Benefits

ACS solvents meet or exceed the high standards of the American Chemical Society (ACS),with test specifications that are specialized to every compound. According to the American Chemical Society, ACS reagent grade implies that it is a substance of sufficient purity to be used in most chemical analyses or reactions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mg2Si nanoparticle synthesis for high pressure hydrogenation.
Chaudhary AL, et al.
The Journal of Physical Chemistry C, 118(2), 1240-1247 (2014)
Practical Synthesis of Di-tert-Butyl-Phosphinoferrocene.
Busacca CA, et al.
Organic Syntheses, 90, 316-326 (2013)
Synthesis of NiO/TiO2 using amphiphilic diblock copolymer brushes (PMMA-b-PAA) by reversible addition fragmentation chain-transfer poltmerization.
Hojjati B and Charpentier PA.
Polymer Reviews, 50(2), 418-418 (2009)
Luca Chiari et al.
The Journal of chemical physics, 138(7), 074301-074301 (2013-03-01)
We present total, elastic, and inelastic cross sections for positron and electron scattering from tetrahydrofuran (THF) in the energy range between 1 and 5000 eV. Total cross sections (TCS), positronium formation cross sections, the summed inelastic integral cross sections (ICS)
Crispin Lichtenberg et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(44), 15797-15805 (2015-09-17)
The reactivity of the all-ferrous FeN heterocubane [Fe4 (Ntrop)4 ] (1) with i) Brønsted acids, ii) σ-donors, iii) σ-donors/π-acceptors, and iv) one-electron oxidants has been investigated (trop = 5H-dibenzo[a,d]cyclo-hepten-5-yl). 1 showed self-re-assembling after reactions with i) and proved surprisingly inert in reactions with ii)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service