Skip to Content
Merck
All Photos(1)

Key Documents

33064

Sigma-Aldrich

2-Methyl-1-propanol

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)

Synonym(s):

Isobutanol, Isobutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
1730878
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
technique(s):
GC/GC: suitable
bp:
108 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

vapor density

2.55 (vs air)

vapor pressure

8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

10.6 %

technique(s)

GC/GC: suitable

impurities

≤0.001% non-volatile matter
≤0.001% peroxides (as H2O2)
≤0.004% free acid (as C3H7COOH)
≤0.005% iso-butyraldehyd (GC)
≤0.005% n-butyraldehyde (GC)
≤0.02% butanone
≤0.05% 2-butanol
≤0.05% water (Karl Fischer)
≤0.1% 1-butanol

refractive index

n20/D 1.3950-1.3960
n20/D 1.396 (lit.)

bp

108 °C (lit.)

mp

−108 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)CO

suitability

passes test for reaction against H2SO4

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1-propanol (Isobutyl alcohol) is widely used organic solvent.

Application

2-Methyl-1-propanol (Isobutyl alcohol) may be used to prepare the isobutyl alcohol-benzene solution for use in the quantification of inorganic phosphates.[1] It may be used for the estimation of phosphate (inorganic phosphorous) by a colorimetric method.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic studies of the catalytic dehydration of isobutyl alcohol on NaH-ZSM-5.
Williams C, et al.
J. Chem. Soc., Faraday Trans., 86(20), 3473-3485 (1990)
Determination of inorganic phosphate.
Martin JB and Doty DM.
Analytical Chemistry, 21(8), 965-967 (1949)
Han Li et al.
Science (New York, N.Y.), 335(6076), 1596-1596 (2012-03-31)
One of the major challenges in using electrical energy is the efficiency in its storage. Current methods, such as chemical batteries, hydraulic pumping, and water splitting, suffer from low energy density or incompatibility with current transportation infrastructure. Here, we report
D Norbäck et al.
Occupational and environmental medicine, 52(6), 388-395 (1995-06-01)
As a part of the worldwide European Community respiratory health survey, possible relations between symptoms of asthma, building characteristics, and indoor concentration of volatile organic compounds (VOCs) in dwellings were studied. The study comprised 88 subjects, aged 20-45 years, from
Pedram Fatehi
Biotechnology progress, 29(2), 297-310 (2013-01-09)
In this review, the recent advancements and technical challenges associated with the production of ethanol, butanol, and isobutanol via bioconversion routes from celluloses of woody materials are reviewed. Physicochemical processes, e.g. steam explosion, seem to be the most viable process

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service