Skip to Content
Merck
All Photos(1)

Key Documents

31247

Sigma-Aldrich

Potassium chlorate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

Assay

≥99%

form

solid

impurities

≤0.0005% heavy metals (as Pb)
≤0.005% in water insoluble matters

mp

356 °C (lit.)

anion traces

bromate (BrO3-): ≤150 mg/kg
chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤20 mg/kg
Fe: ≤3 mg/kg
Mg: ≤10 mg/kg
Na: ≤100 mg/kg

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium chlorate is a shiny colorless solid having monoclinic crystals in the form of platelets. On heating above 400°C, it degrades to give potassium chloride and potassium perchlorate. It has strong oxidizing properties. On exposure to X-rays, oxygen-17 enriched KClO3 generates O3- radical, which was investigated by ESR. On X-ray irradiation potassium chlorate undergoes degradation to generate chloride, chlorite, hypochlorite and occluded oxygen. Significant quantities of dichlorine hexoxide and a trace of chlorine dioxide were also produced.

Application

Potassium chlorate may be used as strong oxidizing agent in the preparation of graphite oxide (GO).

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single sheet functionalized graphene by oxidation and thermal expansion of graphite.
McAllister MJ, et al.
Chemistry of Materials, 19(18), 4396-4404 (2007)
The Decomposition of Solid Potassium Chlorate by X Rays.
Heal HG.
Canadian Journal of Chemistry, 37(5), 979-987 (1959)
ESR Spectrum of O-3 Trapped in a Single Crystal of Potassium Chlorate.
Schlick S.
J. Chem. Phys. , 56, 654-661 (1972)
F Khan et al.
Biochimica et biophysica acta, 1481(2), 229-236 (2000-10-06)
Characterization of conformational transition and folding intermediates is central to the study of protein folding. We studied the effect of various alcohols (trifluoroethanol (TFE), butanol, propanol, ethanol and methanol) and salts (K(3)FeCN(6), Na(2)SO(4), KClO(4) and KCl) on the acid-induced state
Vicente Parra et al.
Physical chemistry chemical physics : PCCP, 9(18), 2266-2273 (2007-05-10)
Here we report on the electroactivity properties of Langmuir-Blodgett (LB) films of the hole-transport molecule 4,4',4''-tris[3-methylphenyl(phenyl)amino] triphenylamine (m-MTDATA). Fairly stable Langmuir films at the air-water interface are accomplished, despite the non-amphiphilic character of the molecule. The reflection-absorption infrared spectroscopy (RAIRS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service