Skip to Content
Merck
All Photos(1)

Key Documents

270660

Sigma-Aldrich

Carbon disulfide

suitable for HPLC, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.9%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
46 °C (lit.)
vapor pressure:
19.48 psi ( 55 °C)

vapor density

2.67 (vs air)

Quality Level

vapor pressure

19.48 psi ( 55 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

212 °F

expl. lim.

50 %

technique(s)

HPLC: suitable

impurities

≤0.03% water

evapn. residue

<0.0005%

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 386 nm Amax: ≤1.0
λ: 388 nm Amax: 0.50
λ: 394 nm Amax: 0.25
λ: 403 nm Amax: 0.10
λ: 410 nm Amax: 0.05
λ: 500-750 nm Amax: 0.01

application(s)

food and beverages

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.
Used as a solvent/reagent to extract:
  • Ethylbenzene, indan, indene and acenaphthene in air samples prior to capillary gas chromatography analysis with flame ionization detection (GC-FID)
  • Hydrocarbons in air prior to GC-MS

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C60: a new form of carbon.
Kratschmer W, et al.
Nature, 347(6291), 354-358 (1990)
Global sources, lifetimes and mass balances of carbonyl sulfide (OCS) and carbon disulfide (CS2) in the earth's atmosphere.
Khalil MAK and Rasmussen RA.
Atmospheric Environment, 18(9), 1805-1813 (1984)
The mass budgets of carbonyl sulfide, dimethyl sulfide, carbon disulfide and hydrogen sulfide.
Watts SF.
Atmospheric Environment, 34(5), 761-779 (2000)
Charles-André Fustin et al.
Langmuir : the ACS journal of surfaces and colloids, 22(15), 6690-6695 (2006-07-13)
We report on a simple procedure to tune the hydrophilicity of hybrid gold nanoparticles. The nanoparticles have been prepared in the core of a poly(ethylene glycol)-block-poly(epsilon-caprolactone) (PEG-b-PCL) five-arm star block copolymer. A hydrophilic corona was then added to these hybrid
Strongly electron deficient sulfonyldithioformate based RAFT agents for hetero Diels-Alder conjugation: Computational design and experimental evaluation.
Nebhani L, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47(22), 6053-6053 (2009)

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service