242241
Activated charcoal
DARCO®, 12-20 mesh, granular
Synonym(s):
Charcoal activated
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
vapor pressure
<0.1 mmHg ( 20 °C)
form
granular
autoignition temp.
842 °F
manufacturer/tradename
DARCO®
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
resistivity
1375 μΩ-cm, 20°C (graphite)
impurities
≤0.40% dust
≤12% moisture content
particle size
12-20 mesh
mp
3550 °C (lit.)
greener alternative category
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.
Application
Activated charcoal (DARCO®, 12-20 mesh, granular) can be used in the effective adsorption of cadmium from aqueous solution. It can also be used in selective adsorption of phenanthrene with a selectivity of ~57.
Other Notes
Acid-washed lignite carbon for general uses.
Legal Information
DARCO is a registered trademark of Cabot Norit Americas, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cadmium and zinc adsorption onto activated carbon: influence of temperature, pH and metal/carbon ratio
Marzal P, et al.
Journal of Chemical Technology and Biotechnology, 66(3), 279-285 (1996)
Cadmium and copper removal by a granular activated carbon in laboratory column systems.
Gabaldon C, et al.
Separation Science and Technology, 35(7), 1039-1053 (2000)
Effect of pH, cation concentration and sorbent concentration on cadmium and copper removal by a granular activated carbon.
Seco A, et al.
Journal of Chemical Technology and Biotechnology, 74(9), 911-918 (1999)
Single and competitive adsorption of Cd and Zn onto a granular activated carbon.
Gabaldon, et al.
Water Research, 30(12), 3050-3060 (1996)
Selective adsorption of phenanthrene dissolved in surfactant solution using activated carbon
Ahn CK, et al.
Chemosphere, 69(11), 1681-1688 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service