227617
Aluminum sulfate octadecahydrate
≥97%
Synonym(s):
Aluminum sulfate hydrate
About This Item
Recommended Products
Quality Level
Assay
≥97%
98.0-102.0% (ACS specification)
form
chunks or granules
crystalline powder
powder or crystals
reaction suitability
reagent type: catalyst
core: aluminum
impurities
≤0.01% insolubles
anion traces
chloride (Cl-): ≤0.005%
cation traces
Ca: 0.01%
Fe: ≤0.002%
K: 0.005%
Mg: 0.002%
Na: 0.02%
heavy metals (as Pb): ≤0.001%
SMILES string
O=S(O)(O)=O.[AlH2][AlH2].O
InChI
1S/2Al.3H2O4S.9H2O/c;;3*1-5(2,3)4;;;;;;;;;/h;;3*(H2,1,2,3,4);9*1H2/q2*+3;;;;;;;;;;;;/p-6
InChI key
KFCWRPFNNIEZRY-UHFFFAOYSA-H
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service