Skip to Content
Merck
All Photos(4)

Key Documents

217638

Sigma-Aldrich

Sodium iodide

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
form:
powder
solubility:
water: soluble(lit.)

vapor density

>1 (vs air)

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder

impurities

<1.0% water

pH

6-9 (20 °C, 50 g/L)

mp

661 °C (lit.)

solubility

water: soluble(lit.)

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium iodide is an ionic compound that can be prepared by treating sodium hydroxide or sodium carbonate with hydriodic acid. It is a versatile reagent used as an iodide source in organic synthesis.

Application

Sodium iodide can be used as a catalyst :
  • To synthesize α-alkoxylated ketones.
  • For N-tert-butoxy carbonylation of amines.
It can also be used as a reagent for the conversion of aryl amines to aryl iodides.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic methods and reactions. 62. Transformations with chlorotrimethylsilane/sodium iodide, a convenient in situ iodotrimethylsilane reagent.
Olah GA, et al.
The Journal of Organic Chemistry, 44(8), 1247-1251 (1979)
Eagleson M.
Concise Encyclopedia Chemistry, 648-648 (1994)
The Chesapeake Bay Institute technique for the Winkler dissolved oxygen method.
Carpenter JH.
Limnology and Oceanography, 10(1), 141-143 (1965)
Eagleson M.
Concise Encyclopedia Chemistry, 999-999 (1994)
Anke de Vries et al.
Investigative radiology, 50(4), 297-304 (2015-01-01)
The objective of this study was to investigate the feasibility and the accuracy of spectral computed tomography (spectral CT) to determine the tissue concentrations and localization of high-attenuation, iodine-based contrast agents in mice. Iodine tissue concentrations determined with spectral CT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service