Skip to Content
Merck
All Photos(1)

Key Documents

15523

Sigma-Aldrich

Glycerol

puriss., anhydrous, 99.0-101.0% (alkalimetric)

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

grade

puriss.

Assay

99.0-101.0% (alkalimetric)
99.0-101.0% (calc. on H2O free substance)

form

viscous liquid

autoignition temp.

698 °F

quality

anhydrous, 99.0-101.0% (alkalimetric)

expl. lim.

2.6-11.3 %

technique(s)

IR spectroscopy: suitable
electrophoresis: suitable

impurities

≤1.0% water (Karl Fischer)

ign. residue

≤0.01% (as SO4)

refractive index

n20/D 1.470-1.475
n20/D 1.474 (lit.)

pH

5.5-8

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.2580-1.2644 g/mL at 20 °C
≥1.257 g/mL at 25 °C
1.25 g/mL (lit.)

cation traces

As: ≤1 mg/kg
Cd: ≤1 mg/kg
Hg: ≤1 mg/kg
Pb: ≤1 mg/kg

suitability

complies for reaction against H2SO4
passes test for identity (IR)

storage temp.

15-25°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol is odourless, colorless, viscous in nature, and exists as a sweet tasting liquid. It can be derived naturally as well as from petrochemical feedstock. Glycerol has a wide variety of applications, and is one of the most valuable and versatile chemical substances in nature. It can be used as an emollient, solvent, sweetening agent, in pharmaceutical formulations, cosmetics, foodstuffs and toiletries. It is very stable and can be easily stored under normal temperature; also it is non-irritating and has no adverse impact to the environment.

Biochem/physiol Actions

Glycerol has been used as
  • a supplement during cell culture of Mycobacterium tuberculosis and Mycobacterium avium.
  • a fuel during designing enzymatic biofuel cell.
  • a liquid composite matrix with 4-HCCA and 3-amino­quinoline for analysis of neutral and acidic glycans.
  • a matrix for fast atom bombardment MS.
  • may be employed as liquid matrix for the quantification studies by MALDI (Matrix-assisted laser desorption/ionization mass spectrometry) analysis.
Glycerol is hygroscopic in nature and is soluble in water owing to its three hydrophilic alcoholic hydroxyl groups. It can form both inter- and intramolecular hydrogen bonds, making it a very flexible molecule. The physiologic effect of glycerine is due to cell-mediated immunity, increased IgG production and increased histamine release.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Hovsep Mahdessian et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(24), 8913-8918 (2014-06-14)
Genome-wide association studies have identified a locus on chromosome 19 associated with plasma triglyceride (TG) concentration and nonalcoholic fatty liver disease. However, the identity and functional role of the gene(s) responsible for these associations remain unknown. Of 19 expressed genes
Benoît Ranchoux et al.
The American journal of pathology, 185(2), 356-371 (2014-12-17)
Pulmonary veno-occlusive disease (PVOD) is an uncommon form of pulmonary hypertension (PH) characterized by progressive obstruction of small pulmonary veins and a dismal prognosis. Limited case series have reported a possible association between different chemotherapeutic agents and PVOD. We evaluated
Marc Simard et al.
The Journal of clinical endocrinology and metabolism, 100(1), E129-E139 (2014-10-17)
Corticosteroid-binding globulin (CBG) is encoded by SERPINA6. It is the major plasma binding protein of glucocorticoids and regulates plasma cortisol levels and bioavailability in humans. Several proteases target CBG and disrupt its steroid-binding properties. To date, most genetic deficiencies that
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service