Skip to Content
Merck
All Photos(2)

Key Documents

154806

Sigma-Aldrich

N,N-Dimethylacetamide

spectrophotometric grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
grade:
spectrophotometric grade
technique(s):
UV/Vis spectroscopy: suitable
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

grade

spectrophotometric grade

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

Assay

≥99%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.050% water

evapn. residue

<0.001%

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 270 nm Amax: 1.00
λ: 280 nm Amax: 0.30
λ: 290 nm Amax: 0.15
λ: 310 nm Amax: 0.05
λ: 320 nm Amax: 0.03
λ: 360-400 nm Amax: 0.01

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide (DMAc) is widely used to compose solvent mixtures for cellulose. Static and dynamic light scattering studies of cellulose dissolved in varying concentrations of DMAc/LiCl mixtures have been reported.

Application

N,N-Dimethylacetamide have been used as solvent in the preparation of nanocomposite materials, comprising of cellulose and hydroxyapatite, using microwave-assisted method. It has also been used as a solvent for poly(ether imides), which can be used to coat magnesium alloys, in order to prevent corrosion.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corrosion protection of magnesium alloy AZ31 by coating with poly (ether imides)(PEI)
Scharnagl.N, et al.
Surface and Coatings Technology, 203(10), 1423-1428 (2009)
Rapid microwave-assisted synthesis and characterization of cellulose-hydroxyapatite nanocomposites in N, N-dimethylacetamide solvent
Ma M-G, et al.
Carbohydrate Research, 345(8), 1046-1050 (2010)
Solutions of cellulose in N, N-dimethylacetamide/lithium chloride studied by light scattering methods.
Roder T, et al.
Polymer, 42(16), 6765-6773 (2001)
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
Wesley Walker et al.
Journal of the American Chemical Society, 135(6), 2076-2079 (2013-01-31)
A major challenge in the development of rechargeable Li-O(2) batteries is the identification of electrolyte materials that are stable in the operating environment of the O(2) electrode. Straight-chain alkyl amides are one of the few classes of polar, aprotic solvents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service