Skip to Content
Merck
All Photos(1)

Key Documents

Y0001556

Ciclesonide impurity C

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ciclesonide

form

powder

packaging

pkg of 10 mg

manufacturer/tradename

EDQM

storage condition

protect from light

solubility

water: <0.1 g/L

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

General description

Ciclesonide impurity C is an impurity of ciclesonide, which is a new-generation, non-halogenated glucocorticoid.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciclesonide impurity C is used as a EP reference standard to quantify the analyte in pharmaceutical formulations using liquid chromatography (LC) technique.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruediger Nave et al.
Clinical pharmacokinetics, 43(7), 479-486 (2004-05-14)
Ciclesonide is a novel inhaled corticosteroid developed for the treatment of asthma. To investigate the extent of oral absorption and bioavailability of ciclesonide referenced to an intravenous infusion. This information provides an estimate for the contribution of the swallowed fraction
New Drugs for Asthma, Allergy and COPD (2001)
Ciclesonide
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 10.3 (2017)
Rethinking Cleaning Validation for API Manufacturing
Zhang C, et al.
Pharmaceutical Technology, 42, 42-54 (2018)
Pharmacokinetics of [14 C] ciclesonide after oral and intravenous administration to healthy subjects
Nave R, et al.
Clinical Pharmacokinetics, 43, 479-486 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service