Skip to Content
Merck
All Photos(1)

Key Documents

Y0001271

Aciclovir for peak identification 1

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

description

API family: acyclovir

API family

acyclovir

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=Nc2c(ncn2COCCO)C(=O)N1

InChI

1S/C8H11N5O3/c9-8-11-6-5(7(15)12-8)10-3-13(6)4-16-2-1-14/h3,14H,1-2,4H2,(H3,9,11,12,15)

InChI key

MKUXAQIIEYXACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aciclovir for peak identification 1 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Prelog et al.
Pediatric nephrology (Berlin, Germany), 26(5), 663-673 (2010-11-16)
Clear recommendations for the management of acute varicella-zoster virus (VZV) infections for cases of significant exposure and the use of prophylactic drugs after solid-organ transplantation are missing due to the lack of evidence by prospective studies. Heterogeneity in patient groups
Todd Hatchette et al.
The Pediatric infectious disease journal, 27(1), 75-77 (2007-12-29)
The authors describe an acyclovir-resistant varicella zoster virus infection in a pediatric patient after hematopoietic stem cell transplant, the use of foscarnet as salvage therapy, and review the literature to clarify the pediatric experience with foscarnet in this setting. A
Suneel Chilukuri et al.
Dermatologic clinics, 21(2), 311-320 (2003-05-22)
In immunocompetent patients, HSV is controlled rapidly by the human host's immune system, and recurrent lesions are small and short lived. When treated with antiviral agents, these patients rarely develop resistance to these drugs. In contrast immunocompromised patients might not
A J Wagstaff et al.
Drugs, 47(1), 153-205 (1994-01-01)
Aciclovir (acyclovir) is a nucleoside analogue with antiviral activity in vitro against the herpes simplex viruses (HSV), varicella zoster virus (VZV), Epstein-Barr virus (EBV), cytomegalovirus (CMV) and human herpesvirus 6 (HHV-6). Topical, oral or intravenous aciclovir is well established in
Jeanne S Sheffield et al.
Obstetrics and gynecology, 102(6), 1396-1403 (2003-12-10)
Genital herpes simplex virus (HSV) infection is one of the most common viral sexually transmitted diseases in the United States. Perinatal transmission of the virus to the fetus or neonate is a major concern in affected pregnancies. Our objective was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service