Skip to Content
Merck
All Photos(1)

Key Documents

Y0001253

Levetiracetam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(αS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, 2(S)-(2-Oxopyrrolidin-1-yl)butyramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levetiracetam

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC[C@H](N1CCCC1=O)C(N)=O

InChI

1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/t6-/m0/s1

InChI key

HPHUVLMMVZITSG-LURJTMIESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levetiracetam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Levetiracetam is a pyrrolidine with antiepileptic activity.
Levetiracetam is a pyrrolidine with antiepileptic activity. Stereoselective binding of levetiracetam was confined to synaptic plasma membranes in the central nervous system with no binding occurring in peripheral tissue. Levetiracetam inhibits burst firing without affecting normal neuronal excitability, which suggests that it may selectively prevent hyper-synchronization of epileptiform burst firing and propagation of seizure activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Consoli et al.
Cerebrovascular diseases (Basel, Switzerland), 34(4), 282-289 (2012-11-07)
Strokes are the leading cause of epileptic seizures in adults and account for 50% of seizures in those over the age of 65 years. The use of antiepileptic drugs to prevent recurrent poststroke seizures is recommended. One hundred and twenty-eight
Hai-Dun Yan et al.
Brain research bulletin, 90, 142-148 (2012-10-31)
Levetiracetam (LEV) is a widely used antiepileptic agent for partial refractory epilepsy in humans. LEV has unique antiepileptic effects in that it does not inhibit electroshock- or pentylenetetrazol-induced convulsion, but does inhibit seizures in kindling animal and spontaneously epileptic rat
Radica M Stepanović-Petrović et al.
Anesthesia and analgesia, 115(6), 1457-1466 (2012-11-13)
We have recently shown that levetiracetam, administered systemically, exerts an antihyperalgesic effect in a rat inflammatory pain model. In this study, we examined whether levetiracetam has local peripheral antihyperalgesic/anti-edematous effects in the same model of localized inflammation and whether opioidergic
Marjolein de Groot et al.
Neuro-oncology, 15(2), 216-223 (2012-12-13)
Treatment of high-grade glioma (HGG) patients with anti-epileptic drugs (AEDs) has met with various side effects, such as cognitive deterioration. The cognitive effects of both older and newer AEDs in HGG patients are largely unknown. The aim of this study
Amy Z Crepeau et al.
Expert review of neurotherapeutics, 10(2), 159-171 (2010-02-09)
Levetiracetam was approved by the US FDA in 1999 after failing the traditional screening tests for antiepileptic drugs. In the 10 years since its introduction, it has become one of the first-line antiepileptic agents and has been evaluated in small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service