Skip to Content
Merck
All Photos(1)

Key Documents

Y0001166

Irbesartan

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Butyl-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4.4]non-1-en-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N6O
CAS Number:
Molecular Weight:
428.53
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

irbesartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC1=NC2(CCCC2)C(=O)N1Cc3ccc(cc3)-c4ccccc4-c5nnn[nH]5

InChI

1S/C25H28N6O/c1-2-3-10-22-26-25(15-6-7-16-25)24(32)31(22)17-18-11-13-19(14-12-18)20-8-4-5-9-21(20)23-27-29-30-28-23/h4-5,8-9,11-14H,2-3,6-7,10,15-17H2,1H3,(H,27,28,29,30)

InChI key

YOSHYTLCDANDAN-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Irbesartan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koen Verdonk et al.
Hypertension (Dallas, Tex. : 1979), 60(3), 722-729 (2012-07-18)
Angiotensin II type 2 (AT(2)) receptor stimulation has been linked to vasodilation. Yet, AT(2) receptor-independent hypertension and hypotension (or no effect on blood pressure) have been observed in vivo after application of the AT(2) receptor agonist compound 21 (C21). We
Ross T Campbell et al.
Journal of the American College of Cardiology, 60(23), 2349-2356 (2012-11-13)
Examination of patients with reduced and preserved ejection fraction in the DIG (Digitalis Investigation Group) trials and the CHARM (Candesartan in Heart Failure: Assessment of Reduction in Mortality and Morbidity) trials provides comparisons of outcomes in each of these types
Zhiling Li et al.
Biomedical chromatography : BMC, 27(2), 254-258 (2012-06-27)
An environmentally friendly ionic liquids dispersive liquid-liquid microextraction (IL-DLLME) method coupled with high-performance liquid chromatography (HPLC) for the determination of antihypertensive drugs irbesartan and valsartan in human urine samples was developed. The HPLC separations were accomplished in less than 10
Optimizing antiplatelet therapy in high-risk patients with atrial fibrillation: insights from Atrial Fibrillation Clopidogrel Trial with Irbesartan for Prevention of Vascular Events (ACTIVE A).
Raymond Chee-Seong Seet et al.
Stroke, 40(12), 3883-3885 (2009-10-31)
Wioleta Marut et al.
Arthritis and rheumatism, 65(5), 1367-1377 (2013-01-22)
Systemic sclerosis (SSc) is characterized by microvascular damage, fibrosis of skin and visceral organs, and autoimmunity. Previous studies have shown that angiotensin II is involved in the synthesis of type I collagen. We investigated whether the blockade of angiotensin II

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service