Skip to Content
Merck
All Photos(1)

Documents

Y0001145

Valsartan for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Valsartan, N-(1-Oxopentyl)-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-L-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N5O3
CAS Number:
Molecular Weight:
435.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

valsartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(=O)N(Cc1ccc(cc1)-c2ccccc2-c3nnn[nH]3)[C@@H](C(C)C)C(O)=O

InChI

1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1

InChI key

ACWBQPMHZXGDFX-QFIPXVFZSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Valsartan for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Teun van der Bom et al.
Circulation, 127(3), 322-330 (2012-12-19)
The role of angiotensin II receptor blockers in patients with a systemic right ventricle has not been elucidated. We conducted a multicenter, double-blind, parallel, randomized controlled trial of angiotensin II receptor blocker valsartan 160 mg twice daily compared with placebo
Moko Zeniya et al.
Hypertension (Dallas, Tex. : 1979), 62(5), 872-878 (2013-09-11)
Na-K-Cl cotransporter isoform 1 (NKCC1) is involved in the regulation of vascular smooth muscle cell contraction. Recently, the with-no-lysine kinase (WNK)-STE20/SPS1-related proline/alanine-rich kinase (SPAK)-NKCC1 phosphorylation cascade in vascular smooth muscle cells was found to be important in the regulation of
Abdul Ahad et al.
International journal of pharmaceutics, 443(1-2), 26-38 (2013-01-15)
The aim of the current investigation is to develop and statistically optimize nanoethosomes for transdermal valsartan delivery. Box-Behnken experimental design was applied for optimization of nanoethosomes. The Independent variables were phospholipids 90G (X(1)), ethanol (X(2)), valsartan (X(3)) and sonication time
Qiuping Ma et al.
International journal of pharmaceutics, 441(1-2), 75-81 (2012-12-26)
The central purpose of this study was to evaluate the impact of drug particle size and crystalline state on valsartan (VAL) formulations in order to improve its dissolution and bioavailability. VAL microsuspension (mean size 22 μm) and nanosuspension (30-80nm) were
Kwang Kon Koh et al.
Diabetes, 62(10), 3547-3552 (2013-07-19)
Statin and angiotensin II type 1 receptor blocker therapy improves endothelial dysfunction using distinct mechanisms. We evaluated simultaneous vascular and metabolic responses to pravastatin and valsartan therapy, alone or in combination, in hypercholesterolemic patients. Forty-eight hypercholesterolemic patients (23 had metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service